Skip to Content
Merck
All Photos(1)

Documents

MAB045P

Sigma-Aldrich

Anti-Fluorescein (FITC) Antibody, clone 5D6.2, Peroxidase conjugated

clone 5D6.2, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

peroxidase conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5D6.2, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable

isotype

IgG2a

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Fluorescein (FITC)

Application

Detect Fluorescein (FITC) using this Anti-Fluorescein (FITC) Antibody, clone 5D6.2, Peroxidase conjugated validated for use in ELISA, IH.
ELISA

Immunohistochemistry

Optimal working dilutions must be determined by end user.

Physical form

0.005M PBS, 50% Stabilzyme, pH7.1

Storage and Stability

Maintain at +2-8°C for up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Reis et al.
The Journal of experimental medicine, 209(9), 1611-1627 (2012-08-22)
Endothelial Wnt/β-catenin signaling is necessary for angiogenesis of the central nervous system and blood-brain barrier (BBB) differentiation, but its relevance for glioma vascularization is unknown. In this study, we show that doxycycline-dependent Wnt1 expression in subcutaneous and intracranial mouse glioma
Amy Barton-Pai et al.
European journal of pharmacology, 657(1-3), 159-166 (2011-02-15)
We tested the hypothesis that glycogen synthase kinase 3α/β (GSK3α/β) modulates tumor necrosis factor-a (TNF) induced increased lung vascular permeability. Rats were treated with TNF (i.v., ~100ng/ml) or vehicle 0.5h, 4.0h and 24.0h prior to lung isolation. Rats were co-treated
Martina Tholen et al.
The Journal of biological chemistry, 290(25), 15758-15769 (2015-05-10)
The cysteine protease cathepsin L (CTSL) is often thought to act as a tumor promoter by enhancing tumor progression and metastasis. This goes along with increased CTSL activity in various tumor entities; however, the mechanisms leading to high CTSL levels

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service