Skip to Content
Merck
All Photos(1)

Documents

KAA002

Sigma-Aldrich

Cellular Senescence Assay

Cellular Senescence Assay Kits provide all the reagents required to efficiently detect SA-β-gal activity at pH 6.0 in cultured cells & tissue sections.

Synonym(s):

Senescence Detection Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

manufacturer/tradename

Chemicon®

technique(s)

cell based assay: suitable

detection method

colorimetric

shipped in

wet ice

General description

Cellular senescence is one of the most fundamental aspects of cell behavior, and is thought to play a critical role in regulating cellular lifespan both in vitro and in vivo [1-3]. Primary somatic cells grown in vitro do not proliferate indefinitely. Instead, after a period of rapid proliferation, cell division rate slows, and ultimately ceases altogether, with the cells becoming unresponsive to mitogenic stimuli. This process is termed cellular senescence, and senescent cells have a well-defined accompanying phenotype - increased cell size, distinctive flat morphology, accumulated lipofuscin granules, wide changes in gene expression, and activity of senescence-associated β-galactosidase (SA-β-gal) [2,3].

It is generally believed that cellular senescence reflects some of the changes that occur during the aging of organisms, and senescent cells have been detected in vivo at sites of age-related pathology, such as benign hyperplastic prostate [4] and atherosclerotic lesions [5]. Recent studies have also provided convincing demonstrations of cellular senescence occurring in vivo in response to internally- and externally-induced stress signals [6,7]. In all of these studies, senescence was characterized by the appearance of senescence-associated β-galactosidase (SA-β-gal) activity, in common with the senescent phenotype in vitro.

Cellular senescence has become an increasingly important target in the development of novel therapeutics. Emerging data implicates senescence bypass in the development of cancer and suggests that senescence may represent a tumor suppressor mechanism. The demonstration that tumor cells can be induced to undergo replicative senescence following the introduction of negative cell-cycle regulators, anti-telomerase peptides, or drug treatment suggests that induction of senescence can be exploited as a basis for cancer therapy [8,9].

For Research Use Only; Not for use in diagnostic procedures

Test Principle:
As outlined above, a classic characteristic of the senescent phenotype is the induction of senescence-associated β-galactosidase (SA-β-gal) activity. SA-β-gal is present only in senescent cells, not in presenescent, quiescent, or proliferating cells. Chemicon′s Cellular Senescence Assay Kit provides all the reagents required to efficiently detect SA-β-gal activity at pH 6.0 in cultured cells and tissue sections. In this assay, SA-β-gal catalyzes the hydrolysis of X-gal, which results in the accumulation of a distinctive blue color in senescent cells. Each kit provides sufficient quantities of reagents to perform at least 50 assays in 35 mm wells.

Application

Cellular Senescence Assay Kits provide all the reagents required to efficiently detect SA-β-gal activity at pH 6.0 in cultured cells & tissue sections.

Components

100X Fixing Solution: (Part No. 2004755) One 1.5 mL vial

10X Staining Solution A: (Part No. 2004756) One 15 mL bottle

10X Staining Solution B: (Part No. 2004754) One 15 mL bottle

X-gal Solution: (Part No. 2004752) Two 1.5 mL vials

Storage and Stability

Store X-gal solution protected from light at -20°C, and other kit components at 4°C. All components supplied are stable for 1 year.

Precautions:
Please refer to the Material Safety Data Sheet at www.chemicon.com for any necessary precautions.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

Flash Point(F)

135.5 °F

Flash Point(C)

57.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard Marcotte et al.
The journals of gerontology. Series A, Biological sciences and medical sciences, 57(7), B257-B269 (2002-06-27)
Forty years after its discovery, replicative senescence remains a rich source of information about cell-cycle regulation and the progression from a normal to a transformed phenotype. Effectors of this growth-arrested state are being discovered at a great pace. This review
E Vasile et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 15(2), 458-466 (2001-02-07)
VPF/VEGF acts selectively on the vascular endothelium to enhance permeability, induce cell migration and division, and delay replicative senescence. To understand the changes in gene expression during endothelial senescence, we investigated genes that were differentially expressed in early vs. late
A Satyanarayana et al.
The EMBO journal, 22(15), 4003-4013 (2003-07-26)
Telomere shortening limits the regenerative capacity of primary cells in vitro by inducing cellular senescence characterized by a permanent growth arrest of cells with critically short telomeres. To test whether this in vitro model of cellular senescence applies to impaired
J Choi et al.
Urology, 56(1), 160-166 (2000-06-28)
Cellular senescence is a unique cellular response pathway thought to be closely associated with the aging process. The senescent phenotype is characterized by the loss of a cell's ability to respond to proliferative and apoptotic stimuli even while normal metabolic
Goberdhan P Dimri
Cancer cell, 7(6), 505-512 (2005-06-14)
Cancer therapeutics are primarily thought to work by inducing apoptosis in tumor cells. However, various tumor suppressors and oncogenes have been shown to regulate senescence in normal cells, and senescence bypass appears to be an important step in the development

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service