Skip to Content
Merck
All Photos(1)

Key Documents

IM1014

Sigma-Aldrich

Anti-Matriptase/MT-SP1 Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-Membrane-Type Serine Protease 1, Anti-Suppressor of Tumorgenicity 14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ST14(6768)

General description

Immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~75 kDa matriptase protein.
Recognizes the ~75 kDa matriptase protein in MCF-7 cells.
This Anti-Matriptase/MT-SP1 Rabbit pAb is validated for use in Immunoblotting, Immunoprecipitation for the detection of Matriptase/MT-SP1.

Immunogen

Human
a synthetic peptide corresponding to amino acids near the C-terminus of human matriptase

Application


Immunoblotting (1:500-1:2000)
Immunoprecipitation (5-20 g/mg lysate)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Highly Toxic (H)

Physical form

In Tris-citrate/phosphate buffer, pH 7.0-8.0.

Reconstitution

Following initial use, aliquot and freeze (-20°C) for long-term storage. Avoid freeze/thaw cycles.

Analysis Note

Positive Control
MCF-7 cells

Other Notes

Antibody should be titrated for optimal results in individual systems.
Santin, A.D., et al. 2003. Cancer98, 1898.
Takeuchi, T., et al. 2000. J. Biol. Chem.275, 26333.
Lin, C.Y., et al. 1999. J. Biol. Chem.274, 18231.
Takeuchi, T., et al. 1999. Proc. Natl. Acad. Sci. USA96, 11054.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sine Godiksen et al.
The Biochemical journal, 413(2), 251-259 (2008-04-12)
HAI-1 [HGF (hepatocyte growth factor) activator inhibitor-1] is a Kunitz-type transmembrane serine protease inhibitor that forms inhibitor complexes with the trypsin-like serine protease, matriptase. HAI-1 is essential for mouse placental development and embryo survival and together with matriptase it is
Ke-Hung Tsui et al.
Journal of molecular endocrinology, 49(2), 153-163 (2012-08-09)
Growth differentiation factor-15 (GDF15), a member of the transforming growth factor-β superfamily, is associated with human cancer progress. We evaluated the role GDF15 plays in tumorigenesis of prostate carcinoma PC-3 cells. Results from real-time RT-PCR and ELISA revealed that expression
Sine Godiksen et al.
PloS one, 8(10), e77146-e77146 (2013-11-10)
Matriptase is a member of the family of type II transmembrane serine proteases that is essential for development and maintenance of several epithelial tissues. Matriptase is synthesized as a single-chain zymogen precursor that is processed into a two-chain disulfide-linked form
Jennifer M Milner et al.
Arthritis and rheumatism, 62(7), 1955-1966 (2010-05-28)
Increasing evidence implicates serine proteinases in pathologic tissue turnover. The aim of this study was to assess the role of the transmembrane serine proteinase matriptase in cartilage destruction in osteoarthritis (OA). Serine proteinase gene expression in femoral head cartilage obtained
Erik W Martin et al.
Oncotarget, 6(32), 33534-33553 (2015-09-24)
The membrane-anchored serine proteases are a unique group of trypsin-like serine proteases that are tethered to the cell surface via transmembrane domains or glycosyl-phosphatidylinositol-anchors. Overexpressed in tumors, with pro-tumorigenic properties, they are attractive targets for protease-activated prodrug-like anti-tumor therapies. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service