Skip to Content
Merck
All Photos(1)

Documents

FCMAB412F

Sigma-Aldrich

Milli-Mark® Anti-Collagen Type I-FITC Antibody, clone 5D8-G9

clone 5D8-G9, Milli-Mark®, from mouse

Synonym(s):

Alpha-1 type I collagen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5D8-G9, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... COL1A1(1277)

General description

Collagens are highly conserved throughout evolution and are characterised by an uninterrupted "Glycine X Y" triplet repeat that is a necessary part of the triple helical structure. Type I collagen (95 kDa) is found in bone, cornea, skin and tendon. Mutations in the encoding gene are associated with osteogenesis imperfecta, Ehlers Danlos syndrome, and idiopathic osteoporosis. Reciprocal translocations between chromosomes 17 and 22, where this gene and the gene for Platelet-derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor.

Specificity

Antibody recognizes Human Collagen Type I.

Immunogen

Purified human type I collagen emulsified in Freund′s complete adjuvant.

Application

Milli-Mark Anti-Collagen Type I-FITC Antibody, clone 5D8-G9 is an antibody against Collagen Type I for use in FC.
Research Category
Cell Structure

Quality

Evaluated by flow cytometry using HEK293T cells.

Target description

139 kDa Calculated

Physical form

Protein A
Purified mouse monoclonal IgG1 conjugated to FITC in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
HEK293T cells

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edmée Eyraud et al.
eLife, 12 (2023-07-26)
Bronchi of chronic obstructive pulmonary disease (COPD) are the site of extensive cell infiltration, allowing persistent contact between resident cells and immune cells. Tissue fibrocytes interaction with CD8+ T cells and its consequences were investigated using a combination of in
Tünde Mester et al.
PloS one, 11(9), e0162994-e0162994 (2016-09-16)
Fibrocytes appear to participate in inflammation and tissue remodeling in patients with thyroid-associated ophthalmopathy (TAO). These patients have increased frequencies of circulating TSH receptor (TSHR)- and CD40-positive fibrocytes, suggesting TSHR and CD40 may play roles in proinflammatory cytokine production, which
Chun-Yu Lo et al.
Respiratory research, 18(1), 194-194 (2017-11-23)
Patients with severe asthma have increased airway remodelling and elevated numbers of circulating fibrocytes with enhanced myofibroblastic differentiation capacity, despite being treated with high doses of corticosteroids, and long acting β2-adrenergic receptor (AR) agonists (LABAs). We determined the effect of
Tanya Wright et al.
Cytometry. Part A : the journal of the International Society for Analytical Cytology, 95(9), 997-1007 (2019-07-10)
Osteosarcoma (OSA) is a malignant tumor of middle-aged dogs and adolescent humans. The clinical outcome of OSA has not improved over more than three decades, and dogs typically succumb to metastatic disease within 6 months despite tumor resection through limb
T Maekawa et al.
Leukemia, 31(12), 2709-2716 (2017-04-08)
Myelofibrosis (MF) may be caused by various pathogenic mechanisms such as elevation in circulating cytokine levels, cellular interactions and genetic mutations. However, the underlying mechanism of MF still remains unknown. Recent studies have revealed that fibrocytes, the spindle-shaped fibroblast-like hematopoietic

Articles

Flow cytometry dye selection tips match fluorophores to flow cytometer configurations, enhancing panel performance.

Flow cytometry dye selection tips match fluorophores to flow cytometer configurations, enhancing panel performance.

Troubleshooting guide offers solutions for common flow cytometry problems, ensuring improved analysis performance.

Troubleshooting guide offers solutions for common flow cytometry problems, ensuring improved analysis performance.

See All

Protocols

Learn key steps in flow cytometry protocols to make your next flow cytometry experiment run with ease.

Explore our flow cytometry guide to uncover flow cytometry basics, traditional flow cytometer components, key flow cytometry protocol steps, and proper controls.

Explore our flow cytometry guide to uncover flow cytometry basics, traditional flow cytometer components, key flow cytometry protocol steps, and proper controls.

Learn key steps in flow cytometry protocols to make your next flow cytometry experiment run with ease.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service