EX0240
Ethyl acetate
for extraction, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
Synonym(s):
Acetic Acid Ethyl Ester, EtOAc
About This Item
for extraction
reagent grade
food and beverages
industrial qc
pharmaceutical
sample preparation
Recommended Products
grade
ACS reagent
for extraction
reagent grade
Quality Level
Agency
meets USP/NF
vapor density
3 (20 °C, vs air)
vapor pressure
73 mmHg ( 20 °C)
description
for analytical purposes
Assay
≥99.5% (GC)
form
liquid
autoignition temp.
801 °F
expl. lim.
2.2-11.5 %, 38 °F
dilution
(for analytical testing)
impurities
≤0.0009 meq/g Titrable acid
≤0.2% Water (H2O)
evapn. residue
≤0.003%
color
APHA: ≤10
refractive index
n20/D 1.3720 (lit.)
bp
76.5-77.5 °C (lit.)
mp
−84 °C (lit.)
transition temp
flash point -4 °C
density
0.902 g/mL at 25 °C (lit.)
application(s)
environmental
food and beverages
industrial qc
pharmaceutical
sample preparation
format
neat
shipped in
ambient
storage temp.
room temp
SMILES string
CCOC(C)=O
InChI
1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3
InChI key
XEKOWRVHYACXOJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Packaging
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!
Analysis Note
Color (APHA): 10 max
Evaporation residue: 0.003% max
Form: Clear liquid
Substances darkened by sulfuric acid: To pass test
Titrable acid: 0.0009meq/g max
Water (H2O): 0.2% max
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
24.8 °F - closed cup
Flash Point(C)
-4 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service