Skip to Content
Merck
All Photos(1)

Key Documents

EX0241

Supelco

Ethyl acetate

OmniSolv®

Synonym(s):

Acetic Acid Ethyl Ester, EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
MDL number:
UNSPSC Code:
12352108
EC Index Number:
205-500-4
technique(s):
HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable
application(s):
PFAS testing
bp:
76.5-77.5 °C (lit.)
vapor pressure:
73 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

3 (20 °C, vs air)

Quality Level

vapor pressure

73 mmHg ( 20 °C)

product line

OmniSolv®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.02% Water (H2O)
≤0.8 μeq/g Titrable acid
≤2 ng/L Capillary ECD responsive substances (as PCNB)

evapn. residue

≤1 ppm

color

APHA: ≤10

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

transition temp

flash point -4 °C

density

0.902 g/mL at 25 °C (lit.)

UV cutoff

≤254 nm

UV absorption

λ: 254 nm Amax: ≤1.00
λ: 255 nm Amax: ≤0.80
λ: 257 nm Amax: ≤0.35
λ: 265 nm Amax: ≤0.02
λ: 270 nm Amax: ≤0.01
λ: 300 nm Amax: ≤0.005

application(s)

PFAS testing

format

neat

shipped in

ambient

storage temp.

room temp

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

EX0241P-1 is packaged in poly coated glass bottles
OmniSolv®

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC)99.9% minCapillary ECD responsive substances (as PCNB)2ng/L maxColor (APHA)10 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFluorescence (as quinine base)250ppt maxFormClear liquidIdentity (IR-spectrum)ConformsRefractive index (at 20°C)Titrable acid0.8µeq/g maxUV Abs. at 254 nm1.00AU maxUV Abs. at 255 nm0.80AU maxUV Abs. at 257 nm0.35AU maxUV Abs. at 265 nm0.02AU maxUV Abs. at 270 nm0.01AU maxUV Abs. at 300 nm0.005AU maxUV Cut-off254nm maxWater (H2O)0.02% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service