CX1054
Chloroform
OmniSolv®
Synonym(s):
Trichloromethane, FM approval, Methylidyne trichloride, Trichloromethane
About This Item
LC/MS: suitable
gas chromatography (GC): suitable
Recommended Products
vapor density
4.1 (vs air)
Quality Level
vapor pressure
160 mmHg ( 20 °C)
product line
OmniSolv®
Assay
≥99.9% (GC)
form
liquid
technique(s)
HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable
impurities
≤0.01% Water (H2O)
≤0.2 μeq/g Titrable acid
≤2 ng/L Capillary ECD responsive substances (as PCNB)
evapn. residue
≤1 ppm
color
APHA: ≤10
refractive index
n20/D 1.445 (lit.)
bp
60.5-61.5 °C (lit.)
mp
−63 °C (lit.)
solubility
8 g/L
density
1.492 g/mL at 25 °C (lit.)
UV cutoff
≤245 nm
UV absorption
λ: 245 nm Amax: ≤1.00
λ: 250 nm Amax: ≤0.30
λ: 260 nm Amax: ≤0.04
λ: 270 nm Amax: ≤0.005
λ: 300 nm Amax: ≤0.005
shipped in
ambient
storage temp.
room temp
SMILES string
ClC(Cl)Cl
InChI
1S/CHCl3/c2-1(3)4/h1H
InChI key
HEDRZPFGACZZDS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Stabilized with Alkene.
Preparation Note
Analysis Note
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3
Target Organs
Central nervous system, Liver,Kidney
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service