Skip to Content
Merck
All Photos(3)

Key Documents

366927

Sigma-Aldrich

Chloroform

suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer

Synonym(s):

Methylidyne trichloride, Trichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein:
1731042
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.04
grade:
HPLC grade
technique(s):
HPLC: suitable
bp:
60.5-61.5 °C (lit.)
vapor pressure:
160 mmHg ( 20 °C)

grade

HPLC grade

Quality Level

vapor density

4.1 (vs air)

vapor pressure

160 mmHg ( 20 °C)

Assay

≥99.8%

form

liquid

purified by

glass distillation

contains

0.5-1.0% ethanol as stabilizer

technique(s)

HPLC: suitable

impurities

Acetone and aldehyde, passes test
Acid and chloride, passes test
Free chlorine (Cl), passes test
H2SO4, passes test (darkened)
≤0.01% water

evapn. residue

<0.0003%

color

APHA: ≤10

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

density

1.48 g/mL at 25 °C
1.492 g/mL at 25 °C (lit.)

cation traces

Pb: ≤0.05 ppm

λ

H2O reference

UV absorption

λ: 245 nm Amax: 1.00
λ: 255 nm Amax: 0.15
λ: 260 nm Amax: 0.15
λ: 270 nm Amax: 0.02
λ: 290 nm Amax: 0.01
λ: 400 nm Amax: 0.01

suitability

passes test for HPLC

storage temp.

room temp

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Novel Approaches for the Analysis and Isolation of Benzylisoquinoline Alkaloids in Chelidonium majus.: This study presents innovative methodologies using chloroform in the extraction and analysis processes, highlighting advances in alkaloid chemistry (Ganzera et al., 2024).

  • Antioxidant, anti-amylase, anti-lipase, and efficiency of Satureja fatty acid on the anti-inflammatory parameters in lipopolysaccharide-stimulated macrophage through Nrf2/NF-kB/NADH oxidase pathway.: Although primarily focused on biological assays, this paper includes essential methodologies employing chloroform for the extraction and purification of biochemical compounds (Saharkhiz MJ et al., 2024).

  • Recent additions and access to a multidimensional lipidomic database containing liquid chromatography, ion mobility spectrometry, and tandem mass spectrometry information.: Chloroform is utilized as a key solvent in the extraction and analysis processes within lipidomics, demonstrating its critical role in modern analytical techniques (Baker ES et al., 2024).

  • Preparation and optimization of dummy molecularly imprinted polymer-based solid-phase extraction system for selective enrichment of p-toluene sulfonate esters genotoxic impurities.: The study includes the use of chloroform in the preparation of polymers for advanced chromatographic applications, illustrating its versatility in analytical methodologies (Shan G et al., 2024).



Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radovan Spurny et al.
The Journal of biological chemistry, 288(12), 8355-8364 (2013-02-01)
Pentameric ligand-gated ion channels (pLGICs), such as nicotinic acetylcholine, glycine, γ-aminobutyric acid GABA(A/C) receptors, and the Gloeobacter violaceus ligand-gated ion channel (GLIC), are receptors that contain multiple allosteric binding sites for a variety of therapeutics, including general anesthetics. Here, we
Philip Jermann et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(33), E3415-E3421 (2014-08-06)
Trimethylation of histone H3 at lysine 27 (H3K27me3) is a chromatin mark associated with Polycomb-mediated gene repression. Despite its critical role in development, it remains largely unclear how this mark is targeted to defined loci in mammalian cells. Here, we
Dominique Bergonier et al.
Veterinary research, 45, 97-97 (2014-10-16)
Staphylococcus aureus is one of the main etiological agents of mastitis in ruminants. In the present retrospective study, we evaluated the potential interest of a previously described automated multiple loci Variable Number of Tandem Repeats (VNTR) Assay (MLVA) comprising 16
Mette Bohl et al.
The American journal of clinical nutrition, 101(4), 870-878 (2015-04-04)
Abdominal obesity and exaggerated postprandial lipemia are independent risk factors for cardiovascular disease (CVD) and mortality, and both are affected by dietary behavior. We investigated whether dietary supplementation with whey protein and medium-chain saturated fatty acids (MC-SFAs) improved postprandial lipid
Kuan Sun et al.
Nature communications, 6, 6013-6013 (2015-01-15)
Solution-processed organic photovoltaic cells (OPVs) hold great promise to enable roll-to-roll printing of environmentally friendly, mechanically flexible and cost-effective photovoltaic devices. Nevertheless, many high-performing systems show best power conversion efficiencies (PCEs) with a thin active layer (thickness is ~100 nm) that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service