Skip to Content
Merck
All Photos(1)

Documents

AP132B

Sigma-Aldrich

Goat Anti-Rabbit IgG Antibody, biotin-SP conjugate

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

biotin conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rabbit

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Affinity purified goat anti-rabbit IgG, Biotin conjugated.
Immunoglobulin G (IgG), is one of the most abundant proteins in human serum with normal levels between 8-17 mg/mL in adult blood. IgG is important for our defence against microorganisms and the molecules are produced by B lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Application

EIA: 1:50,000-1:100,000

Western blots using enzyme-conjugated streptavidin: 1:10,000-1:100,000.

Immunohistochemistry: 1:500-1:5,000

Flow cytometry: 1:200-1:1,000

Fluorescence Immunohisto/cytochemistry: 1:200-1:1,000

Optimal working dilutions must be determined by end user.
Goat anti-Rabbit IgG Antibody, biotin-SP conjugate detects level of Rabbit IgG & has been published & validated for use in ELISA & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Physical form

ImmunoAffinity Purified
RECONSTITUTION:

Reconstitute with sterile distilled water to match the volume indicated on the vial label.

Storage and Stability

Maintain lyophilized product at 2-8°C for up to 12 months. After reconstitution the product is stable for several weeks at 2-8°C as an undiluted liquid. For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of protein (and buffer salts) will decrease to one-half of the original after the addition of glycerol. Avoid repeated freeze/thaw cycles.

WARNING:

For research use only; not for use as a diagnostic.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neuropeptide S facilitates mice olfactory function through activation of cognate receptor-expressing neurons in the olfactory cortex.
Shao, YF; Zhao, P; Dong, CY; Li, J; Kong, XP; Wang, HL; Dai, LR; Hou, YP
Testing null
Laura Saenz del Burgo et al.
The Journal of comparative neurology, 510(6), 581-606 (2008-08-14)
G-protein inwardly rectifying potassium (GIRK) channels mediate the synaptic actions of numerous neurotransmitters in the mammalian brain and play an important role in the regulation of neuronal excitability in most brain regions through activation of various G-protein-coupled receptors such as
Yan Yang et al.
Hypertension (Dallas, Tex. : 1979), 61(2), 519-525 (2012-12-13)
Chronic hypertension is associated with an impaired vascular relaxation caused by an increased vascular tone; however, the underlying mechanisms are not fully understood in human patients. The present study was to investigate whether large-conductance Ca(2+)- and voltage-activated K(+) (BK(Ca)) channels
Detection of anti-tat antibodies in CSF of individuals with HIV-associated neurocognitive disorders.
Bachani, M; Sacktor, N; McArthur, JC; Nath, A; Rumbaugh, J
Journal of Neurovirology null
David Lindenbach et al.
Brain research, 1537, 327-339 (2013-09-26)
Motor symptoms of Parkinson's disease are commonly treated using l-DOPA although long-term treatment usually causes debilitating motor side effects including dyskinesias. A putative source of dyskinesia is abnormally high levels of phosphorylated extracellular-regulated kinase (pERK) within the striatum. In animal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service