Skip to Content
Merck
All Photos(1)

Key Documents

8.51047

Sigma-Aldrich

Dimethyl-N,N-diisopropylphosphoramidite

Novabiochem®

Synonym(s):

Dimethyl-N,N-diisopropylphosphoramidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H20NO2P
CAS Number:
Molecular Weight:
193.22
MDL number:
UNSPSC Code:
12352116

product line

Novabiochem®

Assay

≥95.0% (GC)

form

liquid

reaction suitability

reaction type: Phosphorylations

manufacturer/tradename

Novabiochem®

bp

54 °C/10 hPa

density

1.05 g/cm3

application(s)

peptide synthesis

storage temp.

15-25°C

InChI

1S/C8H20NO2P/c1-7(2)9(8(3)4)12(10-5)11-6/h7-8H,1-6H3

InChI key

KXUMNSXPAYCKPR-UHFFFAOYSA-N

General description

The product number for this product was previously 01-60-0030.

To obtain a certificate of analysis (CoA) of a lot that begins with the letter “A”, please select the option in the right hand menu “Request a COA for Lot#s starting with A”.

Linkage

Replaces: 01-60-0030

Analysis Note

Color (visual): colorless to lightly yellow
Appearance of substance (visual): liquid
Assay (GC, area%): ≥ 95.0 % (a/a)

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(C)

100 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service