Skip to Content
Merck
All Photos(1)

Key Documents

8.03279

Sigma-Aldrich

1,3-Dibromopropane

for synthesis

Synonym(s):

1,3-Dibromopropane, Trimethylene bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)3Br
CAS Number:
Molecular Weight:
201.89
MDL number:
UNSPSC Code:
12352101
EC Index Number:
203-690-3
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

2.6 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

potency

315 mg/kg LD50, oral (Rat)

bp

166-168 °C/1013 hPa

mp

-34 °C

transition temp

flash point 54 °C

density

1.98 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

BrCCCBr

InChI

1S/C3H6Br2/c4-2-1-3-5/h1-3H2

InChI key

VEFLKXRACNJHOV-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.976 - 1.980
Identity (IR): passes test

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service