112216
4-Chlorobenzaldehyde
97%
Synonym(s):
p-Chlorobenzaldehyde
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
ClC6H4CHO
CAS Number:
Molecular Weight:
140.57
Beilstein:
385858
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39050405
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
solid
bp
213-214 °C (lit.)
mp
45-50 °C (lit.)
functional group
aldehyde
chloro
SMILES string
[H]C(=O)c1ccc(Cl)cc1
InChI
1S/C7H5ClO/c8-7-3-1-6(5-9)2-4-7/h1-5H
InChI key
AVPYQKSLYISFPO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
4-Chlorobenzaldehyde reacts with 3-tert-butyl-N-4-chlorobenzyl-1-phenyl-1H-pyrazol-5-amine to form (E)-3-tert-Butyl-4-(4-chlorobenzyl)-N-(4-chlorobenzylidene)-1-phenyl-1H-pyrazol-5-amine by a microwave-mediated reaction.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
197.6 °F - closed cup
Flash Point(C)
92 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
M Reza Naimi-Jamal et al.
Molecular diversity, 14(3), 473-477 (2010-04-08)
Solvent-free one-pot synthesis of 2-amino-4H-chromene scaffold is described in a very simple, efficient, and environmentally benign method using sodium carbonate as a cheap and non-toxic catalyst with up to excellent yields.
Divya P Narayanan et al.
Journal of colloid and interface science, 520, 70-80 (2018-03-13)
A one pot synthesis of carbon dot incorporated porous coconut shell char derived sulphonated catalyst is reported here for the first time and is effectively used in the multicomponent synthesis of amidoalkyl naphthol. Macroporous nature of the char is revealed
Willsingh Anbu Durai et al.
Journal of fluorescence, 30(2), 275-289 (2020-01-31)
Colorimetric sensors have attracted wide scope of attentions due to its fascinating advantages, like handy, equipment-free and naked eye detections. In this investigation, a new and novel hydrazone based dual-responsive ratiometric/colorimetric chemosensor have been developed for highly selective and sensitive
Mohammed M Rahman et al.
Designed monomers and polymers, 21(1), 82-98 (2018-05-31)
A new category of thermally stable hybrid polyarylidene(azomethine-ether)s and copolyarylidene(azomethine-ether)s (PAAP) based on diarylidenecycloalkanones has been synthesized using solution polycondensation method. For potential cationic sensor development, a thin layer of PAAP onto a flat glassy carbon electrode (GCE, surface area:
[Experimental substantiation of an approximate safe level of p-chlorobenzaldehyde in the air of the work area].
A I Khalepo et al.
Gigiena truda i professional'nye zabolevaniia, (2)(2), 50-51 (1987-02-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service