Skip to Content
Merck
All Photos(1)

Key Documents

8.02540

Sigma-Aldrich

Cobalt(II) chloride anhydrous

for synthesis

Synonym(s):

Cobalt(II) chloride anhydrous, Cobalt dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2Co
CAS Number:
Molecular Weight:
129.84
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-589-4
NACRES:
NA.22
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

form

powder

potency

418 mg/kg LD50, oral (Rat)

mp

735 °C

solubility

soluble 585.9 g/L

density

3.36 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

[Co+2].[Cl-].[Cl-]

InChI

1S/2ClH.Co/h2*1H;/q;;+2/p-2

InChI key

GVPFVAHMJGGAJG-UHFFFAOYSA-L

Application

Cobalt(II) chloride anhydrous (CoCl2) can be used as a catalyst in organic synthesis for the transformation of the various functional groups. For example, synthesis of esters via cleavage of ethers by acyl chlorides; acylation of alcohols and amines by anhydrides; and regioselective cleavage of oxiranes. It is also used to synthesize mixed acetals by the addition reaction between alcohols and vinyl ethers.[1]
CoCl2 can be used as a catalyst:
  • For the dealkylation of tertiary amines.[1]
  • In the synthesis of thioketones and thioaldehydes via thionation of carbonyl compounds in the presence of bis(trimethylsilyl) sulfide.[1]
  • In the oxidation of aromatic and aliphatic aldehydes to carboxylic acids in the presence of molecular oxygen and acetic anhydride.[1]
  • In the stereoselective opening of epoxides with N-substituted anilines.[1]

CoCl2 can also be used as:
  • A precursor to synthesize magnetic cobalt nanorods via decomposition of the cobalt coordination complex.[2]
  • A catalyst to prepare acid anhydrides by the reaction of acid chlorides with carboxylic acids.[3]
  • A reagent in the carbon-carbon bond-forming reactions involving active methylene compounds.[1]

Analysis Note

Assay (argentometric): ≥ 98.0 %
Water (K. F.): ≤ 2.0 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cobalt (II) chloride catalyzed syntheses of acid anhydrides from acid chlorides
Srivastava RR and Kabalka GW
Tetrahedron Letters, 33(5) (1992)
Synthesis and magnetic properties of Co nanorod superlattices
Wetz F, et al.
Materials Science and Engineering, C, 27(5-8) (2007)
Cobalt (II) Chloride
Periasamy M, et al.
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service