Skip to Content
Merck
All Photos(1)

Key Documents

662015

Sigma-Aldrich

U 18666A

≥95% (HPLC), lyophilized solid, δ8-sterol isomerase inhibitor, Calbiochem®

Synonym(s):

U18666A, 3β-(2-Diethylaminoethoxy)androst-5-en-17-one, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H41NO2 · xHCl
CAS Number:
Molecular Weight:
387.60 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

U18666A, A cell-permeable, amphiphilic amino-steroid that alters intracellular membrane protein trafficking by impairing intracellular biosynthesis and transport of LDL-derived cholesterol.

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Cl-].N(CCOC1CC2(C3C(C4C(CC3)(C(=O)CC4)C)CC=C2CC1)C)(CC)CC.[H+]

InChI

1S/C25H41NO2.ClH/c1-5-26(6-2)15-16-28-19-9-7-18-8-10-20-21-11-12-23(27)24(21,3)14-13-22(20)25(18,4)17-19;/h8,19-22H,5-7,9-17H2,1-4H3;1H

InChI key

ICPRVJHNLABCSA-UHFFFAOYSA-N

General description

A cell-permeable, amphiphilic amino-steroid that alters intracelluar membrane protein trafficking by impairing intracellular biosynthesis and transport of LDL-derived cholesterol. Reported to exert its effects via the inhibition of 2,3-oxidosqualene-lanosterol cyclase activity. Also reported to inhibit the activity of Δ8-sterol isomerase.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Activity of δ8-sterol isomerase
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hall, A.M., et al. 2003. Pigment Cell Res.16, 149.
Kobayashi, T., et al. 1998. Nature392, 193.
Liscum, L. and Faust, J.R., 1989. J. Biol. Chem.264, 11796.
Sexton, R.C., et al. 1983. Biochemistry22, 5687.
Bierkamper, G.G. and Cenedella, R.J., 1978. Brain Res.150, 343.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebekah L Kendall et al.
European journal of cell biology, 102(2), 151310-151310 (2023-03-20)
Silicosis is considered an irreversible chronic inflammatory disease caused by the inhalation of crystalline silica (cSiO2). The cycle of inflammation that drives silicosis and other particle-caused respiratory diseases is mediated by NLRP3 inflammasome activity in macrophages resulting in the release
Xiaoyan Xu et al.
Nature communications, 15(1), 4237-4237 (2024-05-19)
Immune checkpoint inhibition targeting the PD-1/PD-L1 pathway has become a powerful clinical strategy for treating cancer, but its efficacy is complicated by various resistance mechanisms. One of the reasons for the resistance is the internalization and recycling of PD-L1 itself
Meng Lu et al.
Nature methods, 20(4), 569-579 (2023-03-31)
The ability to quantify structural changes of the endoplasmic reticulum (ER) is crucial for understanding the structure and function of this organelle. However, the rapid movement and complex topology of ER networks make this challenging. Here, we construct a state-of-the-art
Yu-Ting Chiang et al.
International journal of molecular sciences, 24(19) (2023-10-14)
Adipocytes store a significant amount of cholesterol and triglycerides. However, whether cholesterol modulates adipocyte function remains largely unknown. We modulated the cholesterol level in adipocytes to examine its effect on the secretion of adiponectin, an important hormone specifically secreted by
Flavia Giamogante et al.
Cells, 11(10) (2022-05-29)
The study of organelle contact sites has received a great impulse due to increased interest in the understanding of their involvement in many disease conditions. Split-GFP-based contact sites (SPLICS) reporters emerged as essential tools to easily detect changes in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service