Skip to Content
Merck
All Photos(1)

Key Documents

442630-M

Sigma-Aldrich

GMBS

Synonym(s):

GMBS, N-γ-Maleimidobutyryloxysuccinimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O6
CAS Number:
Molecular Weight:
280.23
MDL number:
UNSPSC Code:
12161500
NACRES:
NA.24

Assay

≥90% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

storage temp.

−20°C

SMILES string

N2(C(=O)CCC2=O)OC(=O)CCCN1C(=O)C=CC1=O

InChI

1S/C12H12N2O6/c15-8-3-4-9(16)13(8)7-1-2-12(19)20-14-10(17)5-6-11(14)18/h3-4H,1-2,5-7H2

InChI key

PVGATNRYUYNBHO-UHFFFAOYSA-N

General description

Heterobifunctional reagent useful for preparation of conjugates such as secondary antibodies and β-galactosidase for EIA procedures or for immobilizing enzymes on solid supports.
N-γ-maleimidobutyryloxysuccinimide ester (GMBS) is an aliphatic crosslinker for immunotoxins (IT).(1) It is used for the generation of conjugated Poly-γ-D-glutamic acid.

Application

GMBS is suitable for use:
  • as a hetero-bifunctional cross-linking reagent for the immobilization of neural adhesion molecule L1 onto silicon dioxide surface
  • for the crosslinking of biochips with functionalized gold nanoparticles
  • for coating salinized capillary tube prior to neutravidin treatment in biosensor sandwich immunoassay

Warning

Toxicity: Irritant (B)

Reconstitution

Reconstitute to 10 mg/ml in tetrahydrofuran or DMSO. Add 1 ml of this stock solution dropwise, with stirring, to 10 ml of 20 mM phosphate buffer.

Other Notes

Rusin, K.M., et al. 1992. Biosens. Bioelectron. 7, 367.
Kitagawa, T., et al. 1983. J. Biochem. 94, 1160.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biomolecules detection using a silver-enhanced gold nanoparticle-based biochip
<BIG>Liu Y, et al</BIG>
Nanoscale Research Letters, 5, 533-533 (2010)
Detection of water-borne E. coli O157 using the integrating waveguide biosensor
<BIG>Zhu P, et al</BIG>
Biosensors And Bioelectronics, 21, 678-683 (2005)
Surface immobilization of neural adhesion molecule L1 for improving the biocompatibility of chronic neural probes: In vitro characterization
Acta Biomaterialia, 4, 1208-1217 (2008)
Poly-gamma-D-glutamic acid and protective antigen conjugate vaccines induce functional antibodies against the protective antigen and capsule of Bacillus anthracis in guinea-pigs and rabbits
<BIG>Lee DY, et al</BIG>
FEMS Immunology and Medical Microbiology, 57, Lee DY-Lee DY (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service