Skip to Content
Merck
All Photos(1)

Key Documents

94506

Sigma-Aldrich

Tetramethylrhodamine-5-maleimide

suitable for fluorescence, ≥85% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H23N3O5
CAS Number:
Molecular Weight:
481.50
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.32

Assay

≥85% (HPLC)

form

powder

solubility

DMSO: soluble
methanol: soluble

fluorescence

λex 543 nm; λem 575 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

N5(C(=O)C=CC5=O)c1cc(c(cc1)C2=C3CC=C(C=C3Oc4c2ccc(c4)N(C)C)N(C)C)C(=O)O

InChI

1S/C28H25N3O5/c1-29(2)16-5-9-20-23(14-16)36-24-15-17(30(3)4)6-10-21(24)27(20)19-8-7-18(13-22(19)28(34)35)31-25(32)11-12-26(31)33/h5-9,11-15H,10H2,1-4H3,(H,34,35)

InChI key

KGFLZYXDJDOIEE-UHFFFAOYSA-N

Application

Tetramethylrhodamine-5-maleimide has been used:
  • to concentrate the eluate and in labeling to create IRE1LDS112C
  • to derivatize green fluorescent protein (GFP) on Cys48 for experiments with the extrinsic fluorescent probe
  • to incubate the single solvent exposed cys mutant to perform protein labeling

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Labeling reagent for protein thiols for cytofluorescence

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Specific stereochemistry of OP-1074 disrupts estrogen receptor alpha helix 12 and confers pure antiestrogenic activity
Fanning SW, et al.,
Nature Communications, 9(1), 2368-2368 (2018)
Cytofluorescence techniques for the visualization of distinct pools of protein thiols at the single cell level.
Alfonso Pompella et al.
Methods in molecular biology (Clifton, N.J.), 196, 305-312 (2002-08-03)
Effect of the point mutation H148G on GFPmut2 unfolding kinetics by fluorescence spectroscopy
Bosisio C, et al.
Biophysical Chemistry, 157(1-3), 24-32 (2011)
A J-protein co-chaperone recruits BiP to monomerize IRE1 and repress the unfolded protein response
Amin-WN, et al.
Cell, 171(7), 1625-1637 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service