Skip to Content
Merck
All Photos(1)

Key Documents

420127

Sigma-Aldrich

Jasplakinolide

from Jaspis johnstoni, ≥90% (HPLC), liquid, Actin polymerization promotor, Calbiochem

Synonym(s):

InSolution Jasplakinolide, Jaspis johnstoni

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H45BrN4O6
Molecular Weight:
709.67
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Jasplakinolide, Jaspis johnstoni, InSolution, ≥90%, 1 mM in DMSO, mammablian cell mitosis inhibitor, potent inducer of actin polymerization and stabilization

Quality Level

Assay

≥90% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable F-actin probe. This macrocyclic cyclodepsipeptide, isolated from the marine sponge Jaspis johnstoni, is a potent inducer of actin polymerization and stabilization in vitro. It competes with Phalloidin (Cat. No. 516640) for actin binding (Kd = 15 nM) and enhances apoptosis induced by interleukin 2 (IL-2) deprivation. Also displays fungicidal, insecticidal, and anticancer properties.

Biochem/physiol Actions

Cell permeable: no
Kd = 15 nM for actin binding
Primary Target
F-actin
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Physical form

A 1 mM (50 µg/71 µl) solution of Jasplakinolide, Jaspis johnstoni (Cat. No. 420107) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Rosado, J.A., et al. 2000. J. Biol. Chem.275, 7527.
Posey, S.C., and Bierer, B.E. 1999. J. Biol. Chem.274, 4259.
Senderowicz, A.M., et al. 1995. J. Natl. Cancer Inst.87, 46.
Bubb, M.R., et al. 1994. J. Biol. Chem.269, 14869.
Scott, V.R., et al. 1988. Antimicrob. Agents Chemother.32, 1154.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kem A Sochacki et al.
Developmental cell, 56(8), 1131-1146 (2021-04-07)
Clathrin-mediated endocytosis is the primary pathway for receptor and cargo internalization in eukaryotic cells. It is characterized by a polyhedral clathrin lattice that coats budding membranes. The mechanism and control of lattice assembly, curvature, and vesicle formation at the plasma
Anna M Koester et al.
Biomolecules, 12(8) (2022-07-28)
Recent work suggests that Ras small GTPases interact with the anionic lipid phosphatidylserine (PS) in an isoform-specific manner, with direct implications for their biological functions. Studies on PS-Ras associations in cells, however, have relied on immuno-EM imaging of membrane sheets.
Evdokiia Potolitsyna et al.
Communications biology, 7(1), 458-458 (2024-04-16)
Differentiation of adipose progenitor cells into mature adipocytes entails a dramatic reorganization of the cellular architecture to accommodate lipid storage into cytoplasmic lipid droplets. Lipid droplets occupy most of the adipocyte volume, compressing the nucleus beneath the plasma membrane. How
Kelli L Hvorecny et al.
Nature communications, 15(1), 1840-1840 (2024-02-29)
The cytoskeletal protein actin plays a critical role in the pathogenicity of the intracellular parasite, Toxoplasma gondii, mediating invasion and egress, cargo transport, and organelle inheritance. Advances in live cell imaging have revealed extensive filamentous actin networks in the Apicomplexan

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service