Skip to Content
Merck
All Photos(1)

Key Documents

371720

Sigma-Aldrich

Anti-Giα-1-Subunit, Internal (159-168) Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

diluted serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−70°C

target post-translational modification

unmodified

Gene Information

mouse ... Gnai1(14677)

General description

Anti-Giα-1-Subunit, Internal (159-168), rabbit polyclonal, recognizes Giα-1 subunit. Does not cross-react with Giα-2, Giα-3, Goα, or Gsα subunits. It is validated for use in Western blotting.
Rabbit polyclonal antibody supplied as diluted serum. Recognizes Giα-1 subunit.
Recognizes Giα-1 subunit. Does not cross-react with Giα-2, Giα-3, Goα, or Gsα subunits.

Immunogen

a synthetic peptide (LDRIAQPNYI) corresponding to amino acids 159-168 of mouse Giα-1, conjugated to KLH

Application

Immunoblotting (1:1000)

Warning

Toxicity: Standard Handling (A)

Physical form

Serum diluted in 140 mM NaCl, 100 mM potassium phosphate, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Does not cross-react with Giα-2, Giα-3, Goα, or Gsα subunits. The specificity and cross-reactivity were confirmed with lysates from separate cultures of bacteria transfected with the genes for Giα-1, Giα-2, Giα-3, Goα and Gsα. Variables associated with assay conditions will dictate the proper working dilution.
Kumar, R., et al. 1994. J. Mol. Cell. Cardiol.26, 1537.
Mumby, S.M., and Gilman, A.G. 1991. Methods Enzymol.195, 215.
Goldsmith, P.P., et al. 1987. J. Biol. Chem.262, 14683.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroko Ohtsuka et al.
PloS one, 12(1), e0169609-e0169609 (2017-01-11)
Stromal cell-derived factor-1α (SDF-1α)-induced platelet aggregation is mediated through its G protein-coupled receptor CXCR4 and phosphatidylinositol 3 kinase (PI3K). Here, we demonstrate that SDF-1α induces phosphorylation of Akt at Thr308 and Ser473 in human platelets. SDF-1α-induced platelet aggregation and Akt
Yatin M Patel et al.
Blood, 101(12), 4828-4835 (2003-03-01)
We have examined platelet functional responses and characterized a novel signaling defect in the platelets of a patient suffering from a chronic bleeding disorder. Platelet aggregation responses stimulated by weak agonists such as adenosine diphosphate (ADP) and adrenaline were severely
H-D Yan et al.
British journal of pharmacology, 139(3), 605-615 (2003-06-06)
(1) The effects on the whole-cell carbachol-induced muscarinic cationic current (mIcat) of antibodies against the alpha-subunits of various G proteins, as well as the effect of a Gbetagamma subunit, were studied in single guinea-pig ileal smooth muscle cells voltage-clamped at
Takashi Hirakawa et al.
Endocrinology, 144(9), 3872-3878 (2003-08-23)
Using a Leydig tumor cell line (MA-10) transiently transfected with the human lutropin receptor (hLHR) and mutants thereof, we examined the identity of the G proteins activated by the agonist-engaged hLHR-wild type (wt) and by three of its naturally occurring
S M Gifford et al.
The Journal of endocrinology, 182(3), 485-499 (2004-09-08)
While many endothelial cell lines exist, few are of human origin with characteristics close to the parent endothelial cell. We derived a subline (HUVEC-CS) of immortalized human umbilical vein endothelial cells (HUVEC-C) that proliferate in standard growth media and exhibit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service