262012
Deoxyribonucleic Acid, Sodium Salt, Salmon Testes
Purified DNA from salmon testes. Absorption (0.001% solution in phosphate buffer, pH 7.0, 260 nm): ≥0.155.
Synonym(s):
Deoxyribonucleic Acid, Sodium Salt, Salmon Testes
About This Item
Recommended Products
description
Merck USA index - 14, 2907
Sodium content >=5.8%
Quality Level
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
impurities
Protein, none detected
color
off-white to light brown
white to off-white
solubility
aqueous buffer: soluble
water: soluble
shipped in
ambient
storage temp.
2-8°C
SMILES string
[P](=O)(OC2C(OC(C2)N)CO[P](=O)(OC3C(OC(C3)N)CO)O)(OCC1OC(CC1O)N)O
InChI
1S/C15H31N3O13P2/c16-13-1-7(20)11(28-13)5-25-32(21,22)31-9-3-15(18)29-12(9)6-26-33(23,24)30-8-2-14(17)27-10(8)4-19/h7-15,19-20H,1-6,16-18H2,(H,21,22)(H,23,24)
InChI key
AWBASQCACWFTGD-UHFFFAOYSA-N
General description
Warning
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service