1.99005
Buffer solutions
traceable to NIST, traceable to PTB, pH 4.01 (25 °C, phthalate), pH 7.00 (25 °C, phosphate), pH 9.00(borate), Certipur®
Synonym(s):
pH Calibration Buffers
About This Item
Recommended Products
grade
certified reference material
Quality Level
Agency
traceable to NIST
traceable to PTB
product line
Certipur®
pH
4.01 (25 °C, phthalate)
7.00 (25 °C, phosphate)
9.00(borate)
application(s)
environmental
food and beverages
general analytical
industrial qc
pharmaceutical
format
mixture
storage temp.
15-25°C
General description
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.
Application
Features and Benefits
- Measured and qualified at an ISO/IEC 17025 and ISO 17034 accredited laboratory
- Traceable to secondary reference standards from NIST and primary certified reference standards from PTB
- Ready-to-use solutions available in 30 mL sachets
- Accompanied with a detailed and batch-specific certificate of analysis (CoA) for an effective audit
- Product label follows the ISO Guide 31 and contains information about ordering number, article description, lot number, safety data, and expiry date
- Packaging material subjected to comprehensive stability studies as per ISO Guide 35
- pH 4.00 and pH 6.99 buffer CRMs in the pack are accurate upto +/- 0.02 at 25 °C and the pH 9.00 buffer CRM is accurate upto +/-0.03 at 25 °C
Legal Information
recommended
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service