Skip to Content
Merck
All Photos(1)

Key Documents

12657

Sigma-Aldrich

Albumin, Bovine Serum, Fraction V, Crystalline

Synonym(s):

Albumin, Bovine Serum, Fraction V, Crystalline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

Pricing and availability is not currently available.

description

Merck USA index - 14, 8468

Assay

≥99% (cellulose acetate electrophoresis)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

technique(s)

cell culture | mammalian: suitable

impurities

≤5% moisture

solubility

aqueous buffer: 20 mg/mL
water: 20 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

Prepared by cold ethanol extraction (Cohn method) followed by crystallization.
Prepared from bovine plasma by the Cohn original cold ethanol plasma fractionation method, followed by crystallization at low temperatures. Suitable for sensitive applications requiring high albumin purity and low residual levels of globulins and other biochemical activities. Suitable for the preparation of protein and molecular weight standards, and as a blocking agent for nonspecific binding in ELISA and RIA procedures. It is also useful as an antigen carrier protein in immunological studies and monoclonal antibody production, and as a control protein for enzymatic assays, such as protease, peroxidase, phosphatase or nuclease when determining the purity of reagent grade albumins.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Cohn, F.J., et al. 1947. J. Am. Chem. Soc.69, 1753.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda C Camillo-Andrade et al.
Scientific reports, 10(1), 19392-19392 (2020-11-12)
The continuous search for natural products that attenuate age-related losses has increasingly gained notice; among them, those applicable for skin care have drawn significant attention. The bioester generated from the Chenopodium quinoa's oil is a natural-origin ingredient described to produce
Matthieu Pierre Platre et al.
Nature communications, 13(1), 4445-4445 (2022-08-02)
Iron is critical for host-pathogen interactions. While pathogens seek to scavenge iron to spread, the host aims at decreasing iron availability to reduce pathogen virulence. Thus, iron sensing and homeostasis are of particular importance to prevent host infection and part
Joanna Xiuzhu Xu et al.
Nature communications, 13(1), 7313-7313 (2022-11-28)
The orientation adopted by proteins on nanoparticle surfaces determines the nanoparticle's bioactivity and its interactions with living systems. Here, we present a residue-based affinity scale for predicting protein orientation on citrate-gold nanoparticles (AuNPs). Competitive binding between protein variants accounts for
S Mostoufi-Afshar et al.
Iranian journal of veterinary research, 19(4), 262-269 (2019-02-19)
Mycobacterium avium subspecies paratuberculosis (MAP), as an obligate intracellular bacterium, causes paratuberculosis (Johne's disease) in ruminants. Plus, MAP has consistently been isolated from Crohn's disease (CD) lesions in humans; a notion implying possible direct causative effect for MAP in CD
Shogo Matoba et al.
Genes & development, 36(7-8), 483-494 (2022-04-29)
Genomic imprinting regulates parental origin-dependent monoallelic gene expression. It is mediated by either germline differential methylation of DNA (canonical imprinting) or oocyte-derived H3K27me3 (noncanonical imprinting) in mice. Depletion of Eed, an essential component of Polycomb repressive complex 2, results in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service