12-500
HeLa + EGF Stimulated Cell Lysate in Mg2+ Lysis Buffer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105330
eCl@ss:
32011807
NACRES:
NA.42
Recommended Products
biological source
human
Quality Level
form
liquid
manufacturer/tradename
Upstate®
technique(s)
western blot: suitable
shipped in
dry ice
General description
EGF stimulated HeLa cell lysate. The cell lysate was quantitated via BCA assay. This sell lysate demonstrated higher levels of Ras-GTP form present as compared to unstimulated HeLa cell lysates. The assay was performed using the Ras GTPase Activation ELISA Kit (Catalog # 17-424)
Cell Line Description
HeLa
Physical form
100 &micor;g of EGF stimulated HeLa cell lysate, in 40 µL of 1X MLB (Catalog 20-168) containing, 1 &micor;g/mL leupeptin, 1µg/mL aprotinin, 1 µg/mL pepstatin, 1mM PMSF.
Storage and Stability
Stable for 6 months at -80C from date of shipment. For maximum recovery of product, centrifuge the vial prior to removing the cap. Use immediately in assay following thawing as Ras GTP form can rapidly hydrolyze to the inactive Ras GDP form.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Erik Jue et al.
Lab on a chip, 16(10), 1852-1860 (2016-04-29)
This paper evaluates the potential of 3D printing, a semi-automated additive prototyping technology, as a means to design and prototype a sample-to-device interface, amenable to diagnostics in limited-resource settings, where speed, accuracy and user-friendly design are critical components. As a
April A Dukes et al.
Journal of neurochemistry, 106(1), 333-346 (2008-04-04)
In Parkinson's disease, oxidative stress is implicated in protein misfolding and aggregation, which may activate the unfolded protein response by the endoplasmic reticulum (ER). Dopamine (DA) can initiate oxidative stress via H(2)O(2) formation by DA metabolism and by oxidation into
Gillian A Scullion et al.
Journal of Alzheimer's disease : JAD, 34(3), 781-793 (2013-01-11)
A lifestyle rich in physical and mental activities protects against Alzheimer's disease (AD) but the underlying mechanisms are unclear. We have proposed that this is mediated by a stress response and have shown that repeated exposure to novelty stress, which
Christoph E Joosten et al.
Biotechnology progress, 19(3), 739-749 (2003-06-07)
Secreted human placental alkaline phosphatase (SEAP) was produced in a nonengineered Trichoplusia ni insect cell line, Tn-4s, using a recombinant Autographa californica baculovirus expression vector. The effect of culture conditions on SEAP specific yield and glycosylation was studied. When cultured
D V Marinkovic et al.
The Biochemical journal, 165(3), 417-423 (1977-09-01)
Carboxymethylated beta-galactosidase from Escherichia coli was dissociated at 100 degrees C to form carboxymethylated fragments A and B. The mol.wts. of carboxymethylated fragments A and B were determined by gel filtration to be 64300 and 22400 respectively. Sodium dodecyl sulphate/polyacrylamide-gel
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service