Skip to Content
Merck
All Photos(1)

Key Documents

1.05815

Supelco

Magnesium

powder particle size about 0.06-0.3mm

Synonym(s):

Magnesium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mg
CAS Number:
Molecular Weight:
24.31
MDL number:
UNSPSC Code:
12141502
EC Index Number:
231-104-6
NACRES:
NA.21
Assay:
≥98.5% (complexometric)
form:
solid
Pricing and availability is not currently available.

vapor pressure

0.00013 hPa ( 325 °C)

Quality Level

Assay

≥98.5% (complexometric)

form

solid

autoignition temp.

>450 °C

impurities

≤0.05% Substances insoluble in hydrochloric acid

bp

1107 °C/1013 hPa

mp

651 °C

density

1.74 g/cm3 at 20 °C

cation traces

Fe: ≤500 ppm

storage temp.

no temp limit

SMILES string

[Mg+2]

InChI

1S/Mg/q+2

InChI key

JLVVSXFLKOJNIY-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (complexometric): ≥ 98.5 %
Substances insoluble in hydrochloric acid: ≤ 0.05 %
Fe (Iron): ≤ 500 ppm

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Pyr. Sol. 1 - Water-react 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service