Skip to Content
Merck
All Photos(3)

Documents

549649

Sigma-Aldrich

Magnesium oxide

nanopowder, ≤50 nm particle size (BET)

Synonym(s):

Magnesium(II) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgO
CAS Number:
Molecular Weight:
40.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanopowder

particle size

≤50 nm (BET)

mp

2852 °C (lit.)

SMILES string

O=[Mg]

InChI

1S/Mg.O

InChI key

CPLXHLVBOLITMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cytotoxicity of magnesium oxide nanoparticles on human neural cells and fibroblast was studied. It was found that of all the other metal oxide nanoparticles, MgO nanoparticles were least capable in inducing cell death.

Application

Nanoparticles of magnesium oxide may be used in forming Al2(1-x)MgxTi(1+x)O5 ceramic powder. A composite dielectric material may be prepared by loading poly(methylsilsesquioxane) (PMSQ) on MgO nanoparticles. MgO nanoparticles were used to nucleate ice under a certain temperature and pressure conditions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave sintering of Al2(1-x)MgxTi(1+x)O5 ceramics obtained from mixture of nano-sized oxide powders
Lashkari S and Ebadzadeh T
Ceramics International, 40(8), 12669-12674 (2014)
Facile synthesis of poly (methylsilsesquioxane) and MgO nanoparticle composite dielectrics.
Plank NOV, et al.
Journal of Materials Research, 28(11), 1490-1497 (2013)
Exposure to titanium dioxide and other metallic oxide nanoparticles induces cytotoxicity on human neural cells and fibroblasts.
Lai JCK
International journal of nanomedicine, 3(4), 533-533 (2008)
An aerosol chamber investigation of the heterogeneous ice nucleating potential of refractory nanoparticles.
Saunders RW, et al.
Atmospheric Chemistry and Physics, 10(3), 1227-1247 (2010)
Peter Visser et al.
Faraday discussions, 180, 511-526 (2015-05-01)
Lithium carbonate and lithium oxalate were incorporated as leachable corrosion inhibitors in model organic coatings for the protection of AA2024-T3. The coated samples were artificially damaged with a scribe. It was found that the lithium-salts are able to leach from

Articles

Hydrogen is one of the most important resources in providing food, fuel, and chemical products for our everyday life. Sustainable catalytic hydrogen production from bioethanol has gained significant attention in recent years due to globally diminishing fossil fuel supplies, which have necessitated the search for new chemical feedstocks.

Graphene is a unique two-dimensional (2D) structure of monolayer carbon atoms packed into a dense honeycomb crystal that has attracted great interest due to its diverse and fascinating properties.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service