Skip to Content
Merck
All Photos(2)

Key Documents

1.02822

Supelco

Cyclohexane

for spectroscopy Uvasol®

Synonym(s):

Hexahydrobenzene, Hexamethylene, Naphthene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Beilstein:
1900225
MDL number:
UNSPSC Code:
12191504
EC Index Number:
203-806-2
NACRES:
NA.03
Assay:
≥99.9% (GC)
technique(s):
UV/Vis spectroscopy: suitable
bp:
80.7 °C (lit.)
vapor pressure:
168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

vapor density

2.9 (vs air)

Quality Level

vapor pressure

168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

500 °F

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

9 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.005% Water

evapn. residue

≤0.0002%

color

APHA: ≤10

transmittance

208 nm, ≥20%
220 nm, ≥55%
230 nm, ≥80%
240 nm, ≥90%
250 nm, ≥98%

refractive index

n20/D 1.426 (lit.)

kinematic viscosity

1.26 cSt(20 °C)

bp

80.7 °C (lit.)

mp

4-7 °C (lit.)

transition temp

flash point -18 °C

density

0.779 g/mL at 25 °C (lit.)

UV absorption

λ: 208 nm Amax: ≤0.70
λ: 220 nm Amax: ≤0.26
λ: 230 nm Amax: ≤0.10
λ: 240 nm Amax: ≤0.05
λ: 250 nm Amax: ≤0.01

storage temp.

2-30°C

SMILES string

C1CCCCC1

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2

InChI key

XDTMQSROBMDMFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol<TMSYMBOL></TMSYMBOL> solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol<TMSYMBOL></TMSYMBOL> solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified.

Application


  • Synthesis and Characterization of Graft Copolymers with Poly(ε-caprolactone) Side Chain Using Hydroxylated Poly(β-myrcene-co-α-methyl styrene).: This article details the synthesis and characterization of novel graft copolymers, which include cyclohexane derivatives, for potential use in drug delivery systems, showcasing their thermal and chemical stability which is crucial for pharmaceutical applications (Li T et al., 2024).


  • Direct Extraction and Determination of Free Nicotine in Cigarette Smoke.: This study presents a method for the direct extraction and quantitative analysis of nicotine in cigarette smoke using cyclohexane as a solvent, highlighting its efficiency and safety in analytical chemistry applications (Li L et al., 2024).


Analysis Note

Purity (GC): ≥ 99.9 %
Evaporation residue: ≤ 0.0002 %
Water: ≤ 0.005 %
Colour: ≤ 10 Hazen
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Fluorescence (as quinine at 254 nm): ≤ 1.0 ppb
Fluorescence (as quinine at 365 nm): ≤ 1.0 ppb
Transmission (at 208 nm): ≥ 20 %
Transmission (at 220 nm): ≥ 55 %
Transmission (at 230 nm): ≥ 80 %
Transmission (at 240 nm): ≥ 90 %
Transmission (from 250 nm): ≥ 98 %
Absorbance (at 208 nm): ≤ 0.70
Absorbance (at 220 nm): ≤ 0.26
Absorbance (at 230 nm): ≤ 0.10
Absorbance (at 240 nm): ≤ 0.05
Absorbance (from 250 nm): ≤ 0.01
Meet the requirements of "Specific use spectroscopy" according ACS and Reag. Ph Eur

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

UVASOL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Discover the K270 method for analyzing olive oil quality as per EEC Regulation 2568/91. Access detailed protocols and ensure compliance with standards.

Discover the K270 method for analyzing olive oil quality as per EEC Regulation 2568/91. Access detailed protocols and ensure compliance with standards.

Discover the K270 method for analyzing olive oil quality as per EEC Regulation 2568/91. Access detailed protocols and ensure compliance with standards.

Discover the K270 method for analyzing olive oil quality as per EEC Regulation 2568/91. Access detailed protocols and ensure compliance with standards.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service