Skip to Content
Merck
All Photos(1)

Key Documents

07-531

Sigma-Aldrich

Anti-PIR121-1/Sra-1 Antibody

Upstate®, from rabbit

Synonym(s):

Cytoplasmic FMR1-interacting protein 1, Specifically Rac1-associated protein 1, Sra-1, p140sra-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CYFIP1(23191)
mouse ... Cyfip1(20430)

Specificity

PIR121-1/Sra-1 (aka CYFIP1)

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 872-886 (SQEFQRDKQPNAQPQ-C) of human PIR121-1/Sra-1, with a C-terminal cysteine added for conjugation purposes.

Application

Anti-PIR121-1/Sra-1 Antibody is an antibody against PIR121-1/Sra-1 for use in IP & WB.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

routinely evaluated by immunoblot on RIPA homogenates from rat brain

Target description

Mr~130kDa

Physical form

Format: Purified
Protein A purified

Storage and Stability

stable 2 years at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kuangfu Hsiao et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(5), 1564-1576 (2016-02-05)
Copy number variations encompassing the gene encoding Cyfip1 have been associated with a variety of human diseases, including autism and schizophrenia. Here we show that juvenile mice hemizygous for Cyfip1 have altered presynaptic function, enhanced protein translation, and increased levels
The ARP2/3 complex: giving plant cells a leading edge.
Mathur, Jaideep
Bioessays, 27, 377-387 (2005)
E Saller et al.
The EMBO journal, 18(16), 4424-4437 (1999-08-17)
p53 mutants in tumours have a reduced affinity for DNA and a reduced ability to induce apoptosis. We describe a mutant with the opposite phenotype, an increased affinity for some p53-binding sites and an increased ability to induce apoptosis. The
Patricia Kunda et al.
Current biology : CB, 13(21), 1867-1875 (2003-11-01)
In animal cells, GTPase signaling pathways are thought to generate cellular protrusions by modulating the activity of downstream actin-regulatory proteins. Although the molecular events linking activation of a GTPase to the formation of an actin-based process with a characteristic morphology
Rebecca A Nebel et al.
PloS one, 11(1), e0148039-e0148039 (2016-01-30)
Deletions encompassing the BP1-2 region at 15q11.2 increase schizophrenia and epilepsy risk, but only some carriers have either disorder. To investigate the role of CYFIP1, a gene within the region, we performed knockdown experiments in human neural progenitors derived from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service