Skip to Content
Merck
All Photos(1)

Key Documents

07-052

Sigma-Aldrich

Anti-phospho-Phospholamban (Ser16) Antibody

Upstate®, from rabbit

Synonym(s):

cardiac phospholamban

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, canine, rat, rabbit, bovine, pig, human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer16)

Gene Information

human ... PLN(5350)

General description

Phospholamban (PLB) is a 52 amino acid phosphoprotein which regulates the calcium pump of cardiac sarcoplasmic reticulum (SR). PLB is an oligomer of five identical subunits each having a cytoplasmic and transmembrane domain. The cytoplasmic domain (residues 1 to 25) contains the phosphorylation sites and is highly basic and readily cleaved by proteases; whereas the transmembrane domain (residues 25 to 52) is mostly hydrophobic, protease resistant and stabilizes the pentamer. Following adrenergic stimulation of cardiac muscle, PLB is phosphorylated at Ser16 and at Thr17 which is correlated with stimulation of calcium transport activity across the SR membrane and relaxation of cardiac fibers.

Specificity

Predicted to cross-react with human, mouse, rat, rabbit, pig and dog.
Recognizes phosphorylated phospho-lamban (Ser16), Mr 5-25 kDa. Additional non-specific bands may be detected, Mr 75 & 100 kDa, in some lysates.

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 14-25 of human cardiac phospholamban (RA[pS]TIEMPQQAR-C) with an added C-terminal cysteine. The immunizing sequence is identical in mouse, rat, rabbit, pig and dog.

Application

Research Category
Metabolism

Signaling
Research Sub Category
Muscle Physiology

General Post-translation Modification
This Anti-phospho-Phospholamban (Ser16) Antibody is validated for use in WB for the detection of phospho-Phospholamban (Ser16).

Quality

Routinely evaluated by western blot on bovine cardiac preparation that was incubated with PKA.

Western Blot Analysis:
0.5-2 µg/mL of this lot detected phosphorylated phospholamban from a bovine cardiac preparation that was incubated with 10 µg PKA (Catalog # 14-114) for 10 minutes.
DO NOT BOIL CARDIAC PREPARATION.
Boiling may reduce the 25 kDa pentamer into tri-, di- or monomers. In addition, phosphorylation may disrupt the pentamer into dimers and monomers.

Target description

Varies

Physical form

Immunoaffinity Chromatography
Purified rabbit polyclonal IgG in buffer containing 0.014 M phosphate buffer, pH 7.6, 0.175 M NaCl, 0.07% sodium azide and 30% glycerol. Store at -20°C.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variabillity in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Bovine cardiac preparation, treated with PKA.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E J Mayer et al.
Biochemical and biophysical research communications, 267(1), 40-48 (2000-01-07)
Quantitative immunoassays to discriminate and quantitate phospholamban and its phosphorylation states in heart homogenates were developed using known amounts of protein determined by amino acid analysis. Synthetic 1-52 phospholamban, the hydrophilic 1-25 peptide, and 1-25 phosphopeptides containing P-Ser(16), P-Thr(17), and
Single histidine-substituted cardiac troponin I confers protection from age-related systolic and diastolic dysfunction.
Palpant, NJ; Day, SM; Herron, TJ; Converso, KL; Metzger, JM
Cardiovascular Research null
Bhavini B Patel et al.
Toxicological sciences : an official journal of the Society of Toxicology, 133(1), 174-185 (2013-02-19)
Bisphenol A (BPA) is an estrogenizing endocrine disruptor compound of concern. Our objective was to test whether lifelong BPA would impact cardiac structure/function, calcium homeostasis protein expression, and the DNA methylation of cardiac genes. We delivered 0.5 and 5.0 µg/kg/day
Mark Y Jeong et al.
American journal of physiology. Heart and circulatory physiology, 298(6), H1719-H1726 (2010-04-07)
Using neonatal rat ventricular myocytes, we previously reported that the expression of a dominant negative form of the c-Fos proto-oncogene (AFos) inhibited activator protein 1 activity and blocked the induction of the pathological gene profile stimulated by phenylephrine (PE) while
R Croft Thomas et al.
JACC. Basic to translational science, 1(3), 155-167 (2016-07-28)
Translation of preclinical findings could benefit from a simple, reproducible, high throughput human model to study myocardial signaling. Alpha-1A-adrenergic receptors (ARs) are expressed at very low levels in the human heart, and it is unknown if they function. To develop

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service