Skip to Content
Merck
All Photos(1)

Key Documents

06-864

Sigma-Aldrich

Anti-FLIP Antibody, CT

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CFLAR(8837)

Specificity

long form of FLIP

Immunogen

peptide corresponding to amino acids 447-464 [NGYMYDWNSRVSAKEKYY] of human FLIP

Application

Detect FLIP using this Anti-FLIP Antibody, CT validated for use in WB.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

routinely evaluated by immunoblot on Jurkat, A431, Raji, 3T3, CTLL, PC12 or L6 RIPA cell lysates

Target description

55kDa

Physical form

Format: Purified
PBS, pH 7.4, 0.02% sodium azide before the addition of glycerol to 30%
Protein A chromatography

Storage and Stability

2 years at -20°C

Analysis Note

Control
Positive Antigen Control: Catalog #12-303, Jurkat cell lysate.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Predisposition to apoptosis in keratin 8-null liver is related to inactivation of NF-?B and SAPKs but not decreased c-Flip.
Lee, J; Jang, KH; Kim, H; Lim, Y; Kim, S; Yoon, HN; Chung, IK; Roth, J; Ku, NO
Biology Open null
Paola De Cicco et al.
Frontiers in pharmacology, 8, 65-65 (2017-03-16)
Melanoma is the most common form of skin cancer. Given its high mortality, the interest in the search of preventive measures, such as dietary factors, is growing significantly. In this study we tested, in vitro and in vivo, the potential
Phosphatidylinositol 3-kinase/Akt signaling mediates interleukin-6 protection against p53-induced apoptosis in M1 myeloid leukemic cells.
Vesely, DL; Hoffman, B; Liebermann, DA
Oncogene null
Rongbin Ge et al.
Oncotarget, 6(30), 28973-28987 (2015-09-12)
Metformin has emerged as a potential anticancer agent. Here, we demonstrate that metformin plays an anti-tumor role via repressing N-cadherin, independent of AMPK, in wild-type N-cadherin cancer cells. Ectopic-expression of N-cadherin develops metformin-resistant cancer cells, while suppression of N-cadherin sensitizes
Elisabetta Panza et al.
Pigment cell & melanoma research, 28(1), 61-72 (2014-09-11)
In humans, two main metabolic enzymes synthesize hydrogen sulfide (H2 S): cystathionine γ lyase (CSE) and cystathionine β synthase (CBS). A third enzyme, 3-mercaptopyruvate sulfurtransferase (3-MST), synthesizes H2 S in the presence of the substrate 3-mercaptopyruvate (3-MP). The immunohistochemistry analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service