Skip to Content
Merck
All Photos(4)

Key Documents

06-543

Sigma-Aldrich

Anti-FAK Antibody

Upstate®, from rabbit

Synonym(s):

FADK 1, PTK2 protein tyrosine kinase 2, Protein-tyrosine kinase 2, focal adhesion kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, hamster, mouse, rat

should not react with

avian

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PTK2(5747)

General description

Focal adhesion kinase (FAK) is a non receptor protein tyrosine kinase discovered as a substrate for Src and as a key element of integrin signaling. FAK plays a central role in cell spreading, differentiation, migration, cell death and acceleration of the G1 to S phase transition of the cell cycle. FAK regulation includes phosphorylation at multiple tyrosine and serine residues. Phosphorylation of tyrosine generally is associated with positive regulation and growth promotion, however, dephosphorylation at these sites occurs as cells enter mitosis (M-Phase of the cell cycle). In contrast, serine phosphorylation either remains high or is increased as cells enter mitosis and may play a role in focal adhesion disassembly. FAK and it′s phosphorylation states have been implicated in cancer metastasis and tumor cell survival and adhesion-independent growth. Additionally, recent evidence indicates that elevation of FAK activity in human carcinoma cells is associated with increased invasive potential. A central role in tumor formation and progression suggests that FAK is an attractive target for therapeutic intervention.

Immunogen

pGEX-derived fusion protein containing residues 748-1053 of human FAK

Application

Immunocytochemistry: 10 μg/mL of a previous lot showed positive immunostaining for Fak in Ref 52 cells fixed with 3.7% formaldehyde.

Immunoprecipitation: 4 μg of a previous lot immunoprecipitated FAK from 500 μg of murine 3T3/A31 RIPA lysate.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling
This Anti-FAK Antibody is validated for use in WB, IP, IC for the detection of FAK.

Quality

Evaluated by western blot on RIPA lysates from murine 3T3/A31 or human A431 RIPA cell lysate.

Western Blot Analysis: 0.5-2 μg/mL of this antibody detected FAK in RIPA lysates from murine 3T3/A31 and previously in human A431 RIPA cell lysate.

Target description

125 kDa

Linkage

Replaces: 04-591

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal IgG in buffer containing storage buffer (0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl, 0.05% sodium azide). Store at -20°C.

Storage and Stability

Stable for 1 year at from date of receipt.

Analysis Note

Control
Positive Antigen Control: Catalog #12-305, 3T3/A31 lysate. Add 2.5 μL of 2-mercapto-ethanol/100 μL of lysate and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced lysate per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Focal adhesion kinase-dependent regulation of adhesive forces involves vinculin recruitment to focal adhesions.
David W Dumbauld,Kristin E Michael,Steven K Hanks,Andres J Garcia
Biology of the Cell null
Focal adhesion kinase modulates radial glia-dependent neuronal migration through connexin-26.
Valiente, M; Ciceri, G; Rico, B; Marin, O
The Journal of Neuroscience null
c-Yes regulates cell adhesion at the blood-testis barrier and the apical ectoplasmic specialization in the seminiferous epithelium of rat testes.
Xiang Xiao,Dolores D Mruk,Will M Lee,C Yan Cheng
The International Journal of Biochemistry & Cell Biology null
J Grossmann et al.
Gastroenterology, 120(1), 79-88 (2001-02-24)
Purified intestinal epithelial cells die of detachment-induced apoptosis due to loss of cell anchorage during isolation. Anchorage-dependent cells form focal adhesions, sites of enhanced cell-matrix attachment that confer survival signals. Focal adhesion kinase (FAK), a component of the focal adhesion
Joel B Heim et al.
PloS one, 13(7), e0200558-e0200558 (2018-07-13)
Focal adhesion kinase (FAK) is an intensely studied non-receptor tyrosine kinase with roles in cancer and other common human diseases. Despite the large interest in FAK, the in vivo contribution of FAK auto-phosphorylation site tyrosine (Y) 397 to FAK function

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service