Skip to Content
Merck
All Photos(1)

Key Documents

05-740

Sigma-Aldrich

Anti-phospho-ATM (Ser1981) Antibody, clone 10H11.E12

clone 10H11.E12, Upstate®, from mouse

Synonym(s):

A-T, mutated, AT mutated, TEL1, telomere maintenance 1, homolog, ataxia telangiectasia mutated, ataxia telangiectasia mutated (includes complementation groups A, C and D), ataxia telangiectasia mutated protein, human phosphatidylinositol 3-kinase homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

10H11.E12, monoclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pSer1981)

Gene Information

human ... ATM(472)

General description

Ataxia telangiectasia mutated kinase (ATM) and ataxia telangiectasia and Rad3-related kinase (ATR) are related kinases that regulate cell cycle checkpoints and DNA repair. Mutation in the ATM gene results in the autosomal recessive disease ataxia telangiectasia (AT). The identified substrates for ATM are p53, p95/NBS1, MDM2, Chk2, BRCA1, CtIP, 4E-BP1 and Chk1. The essential requirement for the substrates of ATM/ATR is S/TQ. Hydrophobic amino acids at positions -3 and -1, and negatively charged amino acids at position +1 are positive determinants for substrate recognition by these kinases. Positively charged residues surrounding the S/TQ are negative determinants for substrate phosphorylation. The complex phenotype of cells derived from patients with AT suggests that ATM has additional cellular substrates. In unirradiated cells, ATM is present as an inactive homodimer or multimer. Double-stranded breaks in DNA caused by ionizing radiation cause rapid ATM kinase activation through dissociation of this complex and ATM autophosphorylation at Ser1981.

Specificity

Predicted to cross-react with rat based on sequence homology
This antibody recognizes ATM, Mr ~370 kDa. A non-specific protein was also detected, Mr ~ >400 kDa.

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 1974-1988 (SLAFEEG[pS]QSTTISS) of human ATM. The immunizing sequence has 11/12 identical amino acids in mouse and rat.

Application

Immunoprecipitation:
Phosphorylated ATM was immunoprecipitated from irradiated HeLa cells (Figure A, lanes 3 and 4).

Immunocytochemistry:
Foci are detected in irradiated human and mouse fibroblasts. Determined by an independent laboratory.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Use Anti-phospho-ATM (Ser1981) Antibody, clone 10H11.E12 (Mouse Monoclonal Antibody) validated in ICC, IF, IP, WB to detect phospho-ATM (Ser1981) also known as A-T mutated.

Quality

Routinely evaluated by immunoblot on in crude lysates from irradiated HeLa cells.

Western Blot Analysis:
0.5 µg/mL of this lot detected phosphorylated ATM in crude lysates from irradiated HeLa cells.

Target description

~370 kDa

Physical form

Format: Purified
Protein G Purified
Protein G purified mouse IgG in 0.014 M phosphate buffer, pH 7.6, with 0.175 M NaCl, 0.07 % Sodium Azide and 30% glycerol. Liquid at -20°C.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations:
Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Irradiated HeLa cell lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DNA damage response induced by tobacco smoke in normal human bronchial epithelial and A549 pulmonary adenocarcinoma cells assessed by laser scanning cytometry.
Zhao, Hong, et al.
Cytometry. Part A : the Journal of the International Society For Analytical Cytology, 75, 840-847 (2009)
Fluoroquinolones lower constitutive H2AX and ATM phosphorylation in TK6 lymphoblastoid cells via modulation of the intracellular redox status.
Halicka, H Dorota, et al.
Pharmacological Reports, 61, 711-718 null
DNA repair: Damage alert.
Bartek, Jiri and Lukas, Jiri
Nature, 421, 486-488 (2003)
Na Cao et al.
BMC molecular biology, 17(1), 12-12 (2016-05-11)
Cells respond to DNA damage by activating the phosphatidylinositol-3 kinase-related kinases, p53 and other pathways to promote cell cycle arrest, apoptosis, and/or DNA repair. Here we report that protein palmitoylation, a modification carried out by protein acyltransferases with zinc-finger and
Katarina Ochodnicka-Mackovicova et al.
Journal of immunology (Baltimore, Md. : 1950), 197(7), 2918-2929 (2016-08-26)
The recombination activating gene (RAG) 1 and RAG2 protein complex introduces DNA breaks at Tcr and Ig gene segments that are required for V(D)J recombination in developing lymphocytes. Proper regulation of RAG1/2 expression safeguards the ordered assembly of Ag receptors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service