Skip to Content
Merck
All Photos(2)

Key Documents

05-535

Sigma-Aldrich

Anti-IKKβ Antibody, clone 10AG2

clone 10AG2, Upstate®, from mouse

Synonym(s):

I-kappa-B kinase 2, I-kappa-B-kinase beta, Nuclear factor NF-kappa-B inhibitor kinase beta, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta, inhibitor of nuclear factor kappa B kinase beta subunit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10AG2, monoclonal

species reactivity

rat, human, mouse

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Ikbkb(16150)

General description

IKK beta (I-Kappa-B kinase-beta) is a member of the IKK complex which is composed of IKK alpha, IKK beta, IKK gamma and IKAP. Phosphorylation of I-Kappa-B on a serine residue by the IKK complex frees NF-kB from I-Kappa-B and marks it for degradation via ubiquination. IKK beta has been shown to activate NF-kB and phosphorylate IKB alpha and beta. Phosphorylation of 2 sites at the activation loop of IKK beta is essential for activation of IKK by TNF and IL1. Once activated, IKK beta autophosphorylates which in turn decreases IKK activity and prevents prolonged activation of the inflammatory response. Additionally, IKK beta activity can also be regulated by MEKK-1.

Specificity

Other species not tested.
This antibody recognizes IKKβ/IKK2, Mr 87 kDa.

Immunogen

His-tagged, full length human IKKβ/IKK2. Clone: 10AG2.

Application

Anti-IKKβ Antibody, clone 10AG2 detects level of IKKβ & has been published & validated for use in WB.

Quality

Routinely evaluated by western blot on human Jurkat RIPA cell lysate, human A431, HeLa nuclear extract, mouse 3T3/A31 or rat PC-12 RIPA cell lysates.

Western Blot Analysis:
0.5-2 µg/mL of this lot detected IKK/IKK2 in human HeLa cell lysate. A previous lot was tested in human Jurkat RIPA cell lysate, A431, extract, mouse 3T3/A31 and rat PC-12 RIPA cell lysates.

Target description

87 kDa

Physical form

Format: Purified
Purified mouse monoclonal IgG1 in buffer containing PBS and 0.05% sodium azide. Frozen solution.

Analysis Note

Control
Positive Antigen Control: Catalog #12-303, Jurkat cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emmanuel Derudder et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(18), 5065-5070 (2016-04-22)
Although canonical NF-κB signaling is crucial to generate a normal mature B-cell compartment, its role in the persistence of resting mature B cells is controversial. To resolve this conflict, we ablated NF-κB essential modulator (NEMO) and IκB kinase 2 (IKK2)
Siddharth Bakshi et al.
The Biochemical journal, 474(7), 1163-1174 (2017-02-06)
The double-stranded RNA mimetic poly(I:C) and lipopolysaccharide (LPS) activate Toll-like receptors 3 (TLR3) and TLR4, respectively, triggering the activation of TANK (TRAF family member-associated NF-κB activator)-binding kinase 1 (TBK1) complexes, the phosphorylation of interferon regulatory factor 3 (IRF3) and transcription
Crystal structure of inhibitor of ?B kinase ?.
Xu, G; Lo, YC; Li, Q; Napolitano, G; Wu, X; Jiang, X; Dreano, M; Karin, M; Wu, H
Nature null
Serine phosphorylation of insulin receptor substrate 1 by inhibitor kappa B kinase complex.
Gao, Z; Hwang, D; Bataille, F; Lefevre, M; York, D; Quon, MJ; Ye, J
The Journal of Biological Chemistry null
Validation of IKK beta as therapeutic target in airway inflammatory disease by adenoviral-mediated delivery of dominant-negative IKK beta to pulmonary epithelial cells.
Matthew C Catley, Joanna E Chivers, Neil S Holden, Peter J Barnes, Robert Newton
British Journal of Pharmacology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service