Skip to Content
Merck
All Photos(1)

Documents

05-465

Sigma-Aldrich

Anti-GRK 2/3 (βARK 1/2) Antibody, clone C5/1.1

clone C5/1.1, Upstate®, from mouse

Synonym(s):

G-protein coupled receptor kinase 2, adrenergic, beta, receptor kinase 1, beta adrenergic receptor kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

C5/1.1, monoclonal

species reactivity

rat, mouse, bovine, chicken, human, rabbit, pig

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

General description

The beta-adrenergic receptor kinase specifically phosphorylates the agonist-occupied form of the beta-adrenergic and related G protein-coupled receptors. Overall, the ADRBK2 enzyme, also known as GRK3, has 85% amino acid similarity with ADRBK1, with the protein kinase catalytic domain having 95% similarity. The ADRBK2 mRNA is approximately 8 kilobases with a distribution similar to that of ADRBK1. These data suggest the existence of a family of receptor kinases which may serve broadly to regulate receptor function.
GRK2 phosphorylates the beta2 adrenergic receptor. It is a ubiquitous cytosolic enzyme that specifically phosphorylates the activated form of the beta-adrenergic and related G-protein-coupled receptors.

Specificity

This antibody recognizes GRK2/3, Mr 80 kDa. Additional bands at 85 and 50 kDa are also detected when using higher concentrations of the antibody.

Immunogen

Epitope: a.a. 483-485
Partial GST fusion protein corresponding to the GRK3 (βARK2) carboxyl terminus (a.a. 467-688). This immunogen shares homology with GRK2 (βARK1). The minimal epitope recognized by the antibody is a.a. 483-485. Clone C5/1.1.

Application

Detect GRK 2/3 (βARK 1/2) using this Anti-GRK 2/3 (βARK 1/2) Antibody, clone C5/1.1 validated for use in IP & WB.
Immunoprecipitation:
A previous lot of this antibody was used to immunoprecipitate GRK2/3 from transfected cells, as shown by an independent laboratory.
Research Category
Signaling
Research Sub Category
GPCR, cAMP/cGMP & Calcium Signaling

Quality

Routinely evaluated by immunoblot on RIPA lysates from mouse 3T3/A31 fibroblasts, bovine brain cytosol preparation, rat L6 myoblasts, human A431 carcinoma or human HeLa cell lysates.

Western Blot Analysis:
0.1-1 µg/mL of this lot detected GRK2/3 (βARK 1/2) in RIPA lysates from mouse 3T3/A31 fibroblasts. Previous lots of this antibody detected GRK2/3 in bovine brain cytosol preparation, rat L6 myoblasts, human A431 carcinoma and human HeLa cell RIPA lysates.

Target description

80 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl containing 0.05% sodium azide. Frozen solution.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations:
Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Positive Antigen Control: Catalog #12-305, 3T3/A31 lysate. Add 2.5 μL of 2-mercapto-ethanol/100 μL of lysate and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced lysate per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Iacovelli et al.
Molecular pharmacology, 65(5), 1103-1110 (2004-04-23)
We examined the role of G-protein coupled receptor kinase-2 (GRK2) in the homologous desensitization of mGlu4 metabotropic glutamate receptors transiently expressed in human embryonic kidney (HEK) 293 cells. Receptor activation with the agonist l-2-amino-4-phosphonobutanoate (l-AP4) stimulated at least two distinct
Irini Topalidou et al.
PLoS genetics, 13(10), e1007032-e1007032 (2017-10-03)
The NALCN/NCA ion channel is a cation channel related to voltage-gated sodium and calcium channels. NALCN has been reported to be a sodium leak channel with a conserved role in establishing neuronal resting membrane potential, but its precise cellular role
N Canibe et al.
Journal of animal science, 81(8), 2019-2031 (2003-08-21)
The effect of feeding dry feed (DF), nonfermented liquid feed (NFLF), and fermented liquid feed (FLF) to growing pigs on aspects of gastrointestinal ecology and on performance was investigated. Nonfermented liquid feed was prepared by mixing feed and water at
Maria Olmedo et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(50), 20479-20484 (2012-11-28)
Circadian clocks provide a temporal structure to processes from gene expression to behavior in organisms from all phyla. Most clocks are synchronized to the environment by alternations of light and dark. However, many organisms experience only muted daily environmental cycles
Raphael E Bonita et al.
Clinical and translational science, 3(1), 14-18 (2010-05-07)
G protein-coupled receptor kinase 2 (GRK2), which is upregulated in the failing human myocardium, appears to have a role in heart failure (HF) pathogenesis. In peripheral lymphocytes, GRK2 expression has been shown to reflect myocardial levels. This study represents an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service