Skip to Content
Merck
All Photos(2)

Key Documents

04-796

Sigma-Aldrich

Anti-Akt1/PKBα Antibody, clone AW24 | 04-796

clone AW24, from rabbit

Synonym(s):

AKT1 kinase, Protein kinase B, RAC-alpha serine/threonine-protein kinase, rac protein kinase alpha, v-akt murine thymoma viral oncogene homolog 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

AW24, monoclonal

species reactivity

human, rat, mouse

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... AKT1(207)

General description

The serine threonine protein kinase encoded by the AKT1 gene is catalytically inactive in serum starved primary and immortalized fibroblasts. AKT1 and the related AKT2 are activated by platelet derived growth factor. The activation is rapid and specific. In the developing nervous system AKT is a critical mediator of growth factor induced neuronal survival. Survival factors can suppress apoptosis in a transcription independent manner by activating the serine/threonine kinase AKT1, which then phosphorylates and inactivates components of the apoptotic machinery. Multiple alternatively spliced transcript variants have been found for this gene.

Specificity

This antibody recognizes Akt1/PKBα.

Immunogen

Epitope: C-terminus
Sythetic peptide (C-RPHFPQFSYSASGTA) corresponding to the C-terminal (residues 466-480) of rat Akt1/PKBα.

Application

Anti-Akt1 Antibody, clone AW24 detects level of Akt1/PKBα & has been published & validated for use in WB & IP.
Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling

Quality

Evaluated by western blot on RIPA lysates from human HEK293 cells.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Akt1/PKBα in RIPA lysates from human HEK293 cells.

Target description

~60 kDa

Linkage

Replaces: 05-796

Physical form

Cultured supernantant containing 0.05% sodium azide.
Format: Purified

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HEK293 cell lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María de la Fuente-Fernández et al.
Antioxidants (Basel, Switzerland), 9(4) (2020-04-25)
The incidence of metabolic syndrome (MetS) is increasing worldwide which makes necessary the finding of new strategies to treat and/or prevent it. The aim of this study was to analyze the possible beneficial effects of a carob fruit extract (CSAT+®)
Daniel González-Hedström et al.
Antioxidants (Basel, Switzerland), 9(6) (2020-06-07)
Aging is one of the major risk factors for suffering cardiovascular and metabolic diseases. Due to the increase in life expectancy, there is a strong interest in the search for anti-aging strategies to treat and prevent these aging-induced disorders. Both
Supplementation with a New Standardized Extract of Green and Black Tea Exerts Antiadipogenic Effects and Prevents Insulin Resistance in Mice with Metabolic Syndrome.
De la Fuente-Mu??oz, et al.
International Journal of Molecular Sciences, 24 (2023)
María C Burguete et al.
Journal of neuroendocrinology, 31(8), e12751-e12751 (2019-05-28)
Because neuroprotection in stroke should be revisited in the era of recanalisation, the present study analysed the potential neuroprotective effect of the selective oestrogen receptor modulator, bazedoxifene acetate (BZA), in an animal model of diabetic ischaemic stroke that mimics thrombectomy
Ming-Chun Hung et al.
Cell & bioscience, 12(1), 122-122 (2022-08-03)
Constitutive activation of PI3K signaling has been well recognized in a subset of small cell lung cancer (SCLC), the cancer type which has the most aggressive clinical course amongst pulmonary tumors. Whereas cancers that acquire a mutation/copy gain in PIK3CA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service