Skip to Content
Merck
All Photos(2)

Key Documents

T3125

Sigma-Aldrich

Tetrahydrofolic acid

≥65% (when packaged), powder

Synonym(s):

5,6,7,8-Tetrahydropteroyl-L-glutamic acid, THF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23N7O6
CAS Number:
Molecular Weight:
445.43
Beilstein:
9238187
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic

Quality Level

form

powder

concentration

≥65% (when packaged)

color

faintly yellow to light brown

solubility

0.1 M phosphate pH 7.0: soluble-5 mg/mL, clear to slightly hazy, light yellow to brownish-yellow

storage temp.

−20°C

SMILES string

NC1=NC2=C(NC(CNc3ccc(cc3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)CN2)C(=O)N1

InChI

1S/C19H23N7O6/c20-19-25-15-14(17(30)26-19)23-11(8-22-15)7-21-10-3-1-9(2-4-10)16(29)24-12(18(31)32)5-6-13(27)28/h1-4,11-12,21,23H,5-8H2,(H,24,29)(H,27,28)(H,31,32)(H4,20,22,25,26,30)/t11?,12-/m0/s1

InChI key

MSTNYGQPCMXVAQ-KIYNQFGBSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Tetrahydrofolic acid has been used to determine the inhibitory effect of tetrahydrofolate on polymorphonuclear myeloid-derived suppressor cells (PMN-MDSCs). A recent use of tetrahydrofolate (THF) is for studying the activation of riboswitches.
Tetrahydrofolic acid may also be used as a standard to generate calibration curve to quantify the folate types produced by Streptococcus thermophilus in milk cultures using high-performance liquid chromatography (HPLC). It is also suitable for use in the enzymatic assay of formate-tetrahydrofolate ligase from M. extorquens (MeFtfL).

Biochem/physiol Actions

Tetrahydrofolic acid is the parent molecule of the folate derivatives that donate one-carbon units to amino acids, nucleic acids, and lipids. Tetrahydrofolate metabolites participate in the synthesis of purine and pyrimidine and in synthesis and conversion of amino acids

Packaging

Sealed ampule.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aurora Meucci et al.
Food microbiology, 69, 116-122 (2017-09-25)
The ability of folate-producer strains of Streptococcus thermophilus to accumulate folates and the expression of two target genes (folK and folP), involved in the folate biosynthesis, were studied during milk fermentation. An over-expression of folK took place only in the
Prim-O-glucosylcimifugin enhances the antitumour effect of PD-1 inhibition by targeting myeloid-derived suppressor cells
Gao W, et al.
Journal for Immunotherapy of Cancer, 7(1), 231-231 (2019)
Francisco García-Molina et al.
Journal of agricultural and food chemistry, 59(4), 1383-1391 (2011-01-27)
The coenzyme tetrahydrofolic acid is the most rapid suicide substrate of tyrosinase that has been characterized to date. A kinetic study of the suicide inactivation process provides the kinetic constants that characterize it: λ(max), the maximum apparent inactivation constant; r
Avinash C Srivastava et al.
Plant physiology, 155(3), 1237-1251 (2011-01-15)
A recessive Arabidopsis (Arabidopsis thaliana) mutant with short primary roots and root hairs was identified from a forward genetic screen. The disrupted gene in the mutant encoded the plastidial isoform of folylpolyglutamate synthetase (FPGS), previously designated as AtDFB, an enzyme
Daniela Sumczynski et al.
Food chemistry, 264, 386-392 (2018-06-02)
Non-traditional wild rice flakes were analysed for chemical composition, vitamin B compounds, α-tocopherol, mineral and trace elements. Dietary intakes of vitamins, minerals and trace elements were evaluated using FAO/WHO and Institute of Medicine regulations. Wild rice flakes proved to be

Articles

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

Biofiles reviews innovative technologies for cancer research, reflecting the complexity of the disease.

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service