Skip to Content
Merck
All Photos(2)

Documents

04-1550

Sigma-Aldrich

Anti-SC-35 Antibody, clone 1SC-4F11

ascites fluid, clone 1SC-4F11, from mouse

Synonym(s):

Protein PR264, Splicing component, 35 kDa, Splicing factor SC35, splicing factor, arginine/serine-rich 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1SC-4F11, monoclonal

species reactivity

human, mouse, rat

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SRSF2(6427)

General description

SC-35 is also known as Splicing factor SC35 and Splicing factor, arginine/serine-rich 2. This protein is encoded by the SFRS2 gene and is a member of the splicing factor SR family. SC-35 is involved in pre-mRNA splicing and is found in bodies in the nucleus that are highly enriched in poly(A) RNA. These nuclear bodies are referred to as speckles, SC 35 domains, or splicing factor compartments (SFCs). The purpose of these bodies is still widely disputed. Speckles correspond largely to, if not entirely to, ultra structures called inter-chromatin granule clusters (IGCs). SC3-35 is required for pre-mRNA splicing events and is highly involved in spliceosomal interaction and assembly. SC-35 is necessary for assembly of the ATP-dependent splicing complex. It is also necessary for the ATP-dependent communication between pre-mRNA and U1 and U2 snRNPs.

Specificity

This antibody recognizes SC35.

Immunogen

Epitope: Unknown
Ovalbumin-conjugated linear peptide corresponding to human SC35.

Application

Anti-SC-35 Antibody, clone 1SC-4F11 is a Mouse Monoclonal Antibody for detection of SC-35 also known as Protein PR264, Splicing factor SC35 & has been validated in WB, ICC.
Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected SC35 in NIH/3T3 cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins

Quality

Evaluated by Western Blot in A431 cell lysate.

Western Blot Analysis: 1:1,000 dilution of this antibody detected Sc35 on 10 µg of A431 cell lysate.

Target description

~ 35 kDa

Physical form

Unpurified
Unpurified mouse monoclonal IgG1κ preservative-free ascites.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
A431 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph Pangallo et al.
Blood, 135(13), 1032-1043 (2020-01-22)
Genes encoding the RNA splicing factors SF3B1, SRSF2, and U2AF1 are subject to frequent missense mutations in clonal hematopoiesis and diverse neoplastic diseases. Most "spliceosomal" mutations affect specific hotspot residues, resulting in splicing changes that promote disease pathophysiology. However, a
Hanna Kędzierska et al.
International journal of molecular sciences, 17(10) (2016-10-01)
Serine and arginine rich splicing factor 2(SRSF2) belongs to the serine/arginine (SR)-rich family of proteins that regulate alternative splicing. Previous studies suggested that SRSF2 can contribute to carcinogenic processes. Clear cell renal cell carcinoma (ccRCC) is the most common subtype
Yongchao Liu et al.
Cells, 9(4) (2020-04-16)
The ratio control of 4R-Tau/3R-Tau by alternative splicing of Tau exon 10 is important for maintaining brain functions. In this study, we show that hnRNP A1 knockdown induces inclusion of endogenous Tau exon 10, conversely, overexpression of hnRNP A1 promotes
E Werwein et al.
Cell death & disease, 4, e511-e511 (2013-03-02)
B-Myb is a highly conserved member of the Myb transcription factor family that has essential roles in cell-cycle progression. Recent work has suggested that B-Myb is also involved in the cellular DNA-damage response. Here, we have investigated the fate of
Antisense Oligonucleotide Rescue of Deep-Intronic Variants Activating Pseudoexons in the 6-Pyruvoyl-Tetrahydropterin Synthase Gene.
Mart?-nez-Pizarro, et al.
Nucleic Acid Therapeutics, 32, 378-390 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service