Skip to Content
Merck
All Photos(1)

Key Documents

T-910

Supelco

Triazolam solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12Cl2N4
CAS Number:
Molecular Weight:
343.21
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

Cc1nnc2CN=C(c3ccccc3Cl)c4cc(Cl)ccc4-n12

InChI

1S/C17H12Cl2N4/c1-10-21-22-16-9-20-17(12-4-2-3-5-14(12)19)13-8-11(18)6-7-15(13)23(10)16/h2-8H,9H2,1H3

InChI key

JOFWLTCLBGQGBO-UHFFFAOYSA-N

General description

Triazolam is a benzodiazepine drug used as a sedative to treat severe insomnia. The drug is sold under trade names such as Halcion®, Hypam, and Trilam. This Certified Spiking Solution® is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Halcion is a registered trademark of Pharmacia & Upjohn Company
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David J Greenblatt et al.
Xenobiotica; the fate of foreign compounds in biological systems, 42(12), 1163-1169 (2012-07-19)
A citrus breeding program aimed at developing low furanocoumarin (FC) grapefruit cultivars provided 40 grapefruit juice (GFJ) samples containing variable concentrations of FC derivatives, established as being mechanism-based (irreversible) inhibitors of human CYP3A isoforms. The principal inhibitory FCs were identified
Lawrence P Carter et al.
Psychopharmacology, 226(1), 53-63 (2012-10-26)
Several studies have documented impairments in memory processes as a result of ketamine administration; however, few studies have compared the profile of cognitive effects of ketamine to other drugs. The aim of this study was to compare the cognitive effects
David J Greenblatt et al.
The Journal of pharmacy and pharmacology, 63(2), 214-221 (2011-01-18)
Ketoconazole is extensively used as an index inhibitor of cytochrome P450-3A (CYP3A) activity in vitro and in vivo, but the mechanism of ketoconazole inhibition of CYP3A still is not clearly established. Inhibition of metabolite formation by ketoconazole (seven concentrations from
Guoyou Jin et al.
Talanta, 84(3), 644-650 (2011-04-13)
A triazolam-imprinted silica microsphere was prepared by combining a surface molecular-imprinting technique with the sol-gel process. The results illustrate that the triazolam-imprinted silica microspheres provided using γ-aminopropyltriethoxysilane and phenyltrimethoxysilane as monomers exhibited higher selectivity than those provided from γ-aminopropyltriethoxysilane and
Suicide attempt with an overdose of imatinib.
Osamu Iketani et al.
British journal of clinical pharmacology, 74(5), 890-892 (2012-03-23)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service