Skip to Content
Merck
All Photos(1)

Key Documents

P-010

Supelco

Pentobarbital solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

CCCC(C)C1(CC)C(=O)NC(=O)NC1=O

InChI

1S/C11H18N2O3/c1-4-6-7(3)11(5-2)8(14)12-10(16)13-9(11)15/h7H,4-6H2,1-3H3,(H2,12,13,14,15,16)

InChI key

WEXRUCMBJFQVBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentobarbital is a short-acting barbiturate and hypnotic approved for treatment of seizures and preoperative sedation. Also known as pentobarbitone, the sodium salt of pentobarbital is used as an anesthetic in veterinary medicine. This certified solution standard is suitable for GC/MS and LC/MS testing applications in forensic analysis, clinical toxicology or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brandon H Gilroyed et al.
Journal of veterinary science, 14(1), 103-106 (2013-02-08)
In the event of an infectious disease outbreak in cattle, carcasses must be disposed of in a rapid and contained manner. This brief communication details injection of a barbiturate to euthanize cattle inoculated with Escherichia coli O157:H7 followed by carcass
M Katayama et al.
Laboratory animals, 47(2), 122-126 (2013-04-09)
Pharmacokinetics of drugs may differ between small and large mammals (including humans); therefore, drug testing in animal models must be carefully designed. Sprague-Dawley rats were used in cardiac experiments, during which the lopinavir concentration in serum had to match human
Nassir Mansour et al.
Journal of neurosurgery. Pediatrics, 12(1), 37-43 (2013-05-07)
Barbiturates are widely used in the management of high intracranial pressure (ICP) caused by diffuse brain swelling. The cardiovascular, renal, and immunological side effects of these drugs limit them to last-line therapy. There are few published data regarding the role
Maria Pedro Guarino et al.
In vivo (Athens, Greece), 27(1), 127-132 (2012-12-15)
We compared the effects of two different anaesthetics, sodium pentobarbital (65 mg/kg) and ketamine (30 mg/kg) plus xylazine (4 mg/kg) (KX) on insulin sensitivity, fasting glycaemia, insulinaemia and free fatty acids (FFA). Four groups of Wistar rats were used: KX
Sarah A Mellion et al.
Pediatric critical care medicine : a journal of the Society of Critical Care Medicine and the World Federation of Pediatric Intensive and Critical Care Societies, 14(3), 239-247 (2013-02-09)
To evaluate high-dose barbiturates as a second-tier therapy for pediatric refractory intracranial hypertension complicating severe traumatic brain injury. This is a retrospective cohort study of children with refractory intracranial hypertension treated with high-dose barbiturates. A single center level I pediatric

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service