Skip to Content
Merck
All Photos(1)

Key Documents

N-079

Supelco

(R,S)-Norcotinine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10N2O
CAS Number:
Molecular Weight:
162.19
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

certified reference material

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

format

single component solution

storage temp.

−20°C

SMILES string

O=C1NC(C2=CN=CC=C2)CC1

InChI

1S/C9H10N2O/c12-9-4-3-8(11-9)7-2-1-5-10-6-7/h1-2,5-6,8H,3-4H2,(H,11,12)

InChI key

FXFANIORDKRCCA-UHFFFAOYSA-N

General description

Norcotinine, or desmethylcotinine, is a urinary metabolite of nornicotine and cotinine found in smokers and users of other nicotine products including chewing tobacco and snuff. This Certified Spiking Solution® is suitable for use in nicotine testing methods or clinical and diagnostic testing of nicotine biomarkers by LC-MS/MS or GC/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diaa M Shakleya et al.
Analytical and bioanalytical chemistry, 395(7), 2349-2357 (2009-10-20)
An analytical procedure was developed and validated for the simultaneous identification and quantification of nicotine, cotinine, trans-3'-hydroxycotinine, and norcotinine in 0.5 mL of human oral fluid collected with the Quantisal oral fluid collection device. Solid phase extraction and liquid chromatography-tandem
H Foth et al.
Toxicology and applied pharmacology, 105(3), 382-392 (1990-09-15)
Elimination parameters of [14C]nicotine in conscious rats receiving nicotine (0.3 mg/kg) either intravenously or orally were studied. The oral availability of unchanged nicotine, derived by comparison of the respective areas under the concentration vs time curves (AUC), was 89%, indicating
Janne Hukkanen et al.
Clinical pharmacology and therapeutics, 80(5), 522-530 (2006-11-23)
Grapefruit juice is an inhibitor of the cytochrome P450 (CYP) 3A4 enzyme and transporters such as P-glycoprotein and organic anion transporting polypeptides, leading to clinically important interactions. Our objective was to study the effect of grapefruit juice on the pharmacokinetics
Liza U Ljungberg et al.
Endothelium : journal of endothelial cell research, 15(5-6), 239-245 (2008-12-10)
Nicotine has been shown to induce endothelial dysfunction, which is an early marker of atherosclerosis. Nicotine undergoes extensive metabolism in the liver, forming a number of major and minor metabolites. There are very limited data on the effect of nicotine
N Eldirdiri et al.
European journal of drug metabolism and pharmacokinetics, 22(4), 385-390 (1998-03-26)
Since norcotinine and 4-(3-pyridyl)-4-oxobutyramide (POBAM) are probable metabolites of nicotine and cotinine, it was of interest to investigate the further in vitro metabolism of these compounds. We now report our preliminary findings using rat microsomal preparations (induced and/or non-induced with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service