Skip to Content
Merck
All Photos(1)

Key Documents

N-004

Supelco

Naloxone solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21NO4
CAS Number:
Molecular Weight:
327.37
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

Oc1ccc2C[C@H]3N(CC[C@@]45[C@@H](Oc1c24)C(=O)CC[C@@]35O)CC=C

InChI

1S/C19H21NO4/c1-2-8-20-9-7-18-15-11-3-4-12(21)16(15)24-17(18)13(22)5-6-19(18,23)14(20)10-11/h2-4,14,17,21,23H,1,5-10H2/t14-,17+,18+,19-/m1/s1

InChI key

UZHSEJADLWPNLE-GRGSLBFTSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Naloxone is an opiate drug used to counter the effects of opioid overdose. This certified solution standard is applicable for use in LC/MS or GC/MS applications such as clinical toxicology and forensic analysis. Naloxone is sold under the trade names Narcan®, Nalone, and Narcan®ti.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Narcan is a registered trademark of Endo Pharmaceuticals Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Terry Furst
Journal of addictive diseases, 32(1), 53-67 (2013-03-14)
This study explores strategies that Suboxone misusers utilize while in drug treatment. Ethnographic interviews were conducted with 14 patients who had cycled in and out of Suboxone treatment. The objective of the study is to identify strategies implemented by patients
Anne M Neumann et al.
Journal of addictive diseases, 32(1), 68-78 (2013-03-14)
Patients with opioid addiction who receive prescription opioids for treatment of nonmalignant chronic pain present a therapeutic challenge. Fifty-four participants with chronic pain and opioid addiction were randomized to receive methadone or buprenorphine/naloxone. At the 6-month follow-up examination, 26 (48.1%)
Liang-Wu Fu et al.
American journal of physiology. Heart and circulatory physiology, 305(1), H76-H85 (2013-05-07)
Thinly myelinated Aδ-fiber and unmyelinated C-fiber cardiac sympathetic (spinal) sensory nerve fibers are activated during myocardial ischemia to transmit the sensation of angina pectoris. Although recent observations showed that myocardial ischemia increases the concentrations of opioid peptides and that the
Robert Taylor et al.
Expert opinion on investigational drugs, 22(4), 517-525 (2013-03-15)
Opioid receptor antagonists are well known for their ability to attenuate or reverse the effects of opioid agonists. This property has made them useful in mitigating opioid side effects, overdose and abuse. Paradoxically, opioid antagonists have been reported to produce
Severe postoperative hyperthermic syndrome after addition of tilidine/naloxone to duloxetine therapy.
Marco Vinetti et al.
Clinical toxicology (Philadelphia, Pa.), 51(6), 516-517 (2013-06-19)

Articles

Although both biphenyl and phenyl-hexyl phases can resolve these compounds, the former exhibits excellent peak shape and substantially less silanol-derived ion exchange activity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service