Skip to Content
Merck
All Photos(1)

Key Documents

N129

Sigma-Aldrich

Naloxone methiodide

≥98% (HPLC), solid

Synonym(s):

(5α,17R)-4,5-Epoxy-3,14-dihydroxy-17-methyl-6-oxo-17-(2-propenyl)-morphinanium iodide, N-Methylnaloxonium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24INO4
CAS Number:
Molecular Weight:
469.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

color

white to tan

solubility

H2O: >10 mg/mL

storage temp.

2-8°C

SMILES string

[I-].[H][C@]12Oc3c(O)ccc4C[C@@H]5[C@](O)(CCC1=O)[C@@]2(CC[N@+]5(C)CC=C)c34

InChI

1S/C20H23NO4.HI/c1-3-9-21(2)10-8-19-16-12-4-5-13(22)17(16)25-18(19)14(23)6-7-20(19,24)15(21)11-12;/h3-5,15,18,24H,1,6-11H2,2H3;1H/t15-,18+,19+,20-,21?;/m1./s1

InChI key

ICONPJDAXITIPI-UXYWFNEESA-N

Application

Naloxone methiodide has been used as inhibitor of opioid receptor in fish and mice. Naloxone methiodide has been used to block opioid neurotransmission.

Biochem/physiol Actions

Naloxone methiodide has low affinity for opioid receptors than naloxone. Administration of naloxone inhibits opioid receptor and opioid-induced respiratory depression.
Quaternary salt of naloxone that, like the parent compound, is a nonselective antagonist at opioid receptors. It does not cross the blood-brain barrier.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piotr Wojciechowski et al.
European journal of pharmacology, 885, 173514-173514 (2020-08-30)
PK20M (Dmt-D-Lys-Phe-Phe-OH) is a novel modified endomorphin-2 (EM-2) peptide producing strong dose- and time-dependent antinociceptive activity. Yet its prototype, endogenous EM-2, has been reported to trigger respiratory and vascular effects such as apnea and hypotension. The purpose of this study
Delta opioid receptors are essential to the antiallodynic action of Β2-mimetics in a model of neuropathic pain.
Mélanie Kremer et al.
Molecular pain, 16, 1744806920912931-1744806920912931 (2020-03-27)
Naloxone and its quaternary derivative, naloxone methiodide, have differing affinities for mu, delta, and kappa opioid receptors in mouse brain homogenates
Lewanowitsch Tanya and Irvine RJ
Brain Research, 964(2), 302 -3305 (2003)
Helvira Melo et al.
Scientific reports, 8(1), 15551-15551 (2018-10-21)
Opioids remain the gold standard for the treatment of moderate to severe pain. However, their analgesic properties come with important side effects, including pruritus, which occurs frequently after systemic or neuraxial administration. Although part of the opioid-induced itch is mediated
Naloxone methiodide reverses opioid-induced respiratory depression and analgesia without withdrawal
Lewanowitsch Tanya and Irvine RJ
European Journal of Pharmacology, 445(1-2), 61 -667 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service