Skip to Content
Merck
All Photos(1)

Key Documents

N-003

Supelco

Norcocaine hydrochloride solution

1.0 mg/mL in acetonitrile (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19NO4 · HCl
CAS Number:
Molecular Weight:
325.79
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.COC(=O)[C@@H]1[C@H]2CC[C@@H](C[C@@H]1OC(=O)c3ccccc3)N2

InChI

1S/C16H19NO4.ClH/c1-20-16(19)14-12-8-7-11(17-12)9-13(14)21-15(18)10-5-3-2-4-6-10;/h2-6,11-14,17H,7-9H2,1H3;1H/t11-,12+,13-,14+;/m0./s1

InChI key

RIPPXFWUYXTRNJ-BHARVXRSSA-N

General description

Norcocaine is a primary urinary metabolite of the popular recreation drug cocaine. Cocaine is a central nervous system stimulant with appetite suppressant and topical anesthetic properties. This certified solution standard is applicable for use in urine drug testing, clinical toxicology or forensic toxicology applications by GC/MS or LC/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M G Ladona et al.
Life sciences, 68(4), 431-443 (2001-02-24)
Cocaine N-demethylation to norcocaine was studied in human liver microsomes of different ages. Norcocaine was formed at a considerable rate in fetal (45.4+/-18.2 nmol/mg x hour, n = 8) and adult specimens (82.0+/-46.6 nmol/mg x hour, n = 15), p
Christine Moore et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 859(2), 208-212 (2007-10-24)
A quantitative analytical procedure for the determination of cocaine, benzoylecgonine and cocaethylene and norcocaine in hair has been developed and validated. The hair samples were washed, incubated, and any drugs present were quantified using mixed mode solid-phase extraction and liquid
C W Schindler et al.
European journal of pharmacology, 431(1), 53-59 (2001-11-22)
The effects of cocaine and the cocaine metabolites norcocaine, ecgonine methyl ester, benzoylecgonine and cocaethylene were evaluated in conscious squirrel monkeys for their effects on blood pressure and heart rate. Norcocaine, ecgonine methyl ester and benzoylecgonine are produced in vivo
P Pellinen et al.
Archives of toxicology, 74(9), 511-520 (2000-12-29)
The first step in the oxidative metabolism of cocaine is N-demethylation to norcocaine, which is further N-hydroxylated to more toxic N-hydroxynorcocaine. In this study we examined the kinetics of norcocaine N-hydroxylation mediated by cytochrome P450 (CYP) in mouse and human
Peter Kovacic
Medical hypotheses, 64(2), 350-356 (2004-12-21)
Cocaine is one of the principal drugs of abuse. Although impressive advances have been made, unanswered questions remain concerning mechanism of toxicity and addiction. Discussion of action mode usually centers on receptor binding and enzyme inhibition, with limited attention to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service