Skip to Content
Merck
All Photos(1)

Documents

L6533

Sigma-Aldrich

Lectin from Dolichos biflorus (horse gram)

biotin conjugate, buffered aqueous solution

Synonym(s):

Dolichos biflorus agglutinin, DBA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

conjugate

biotin conjugate

Quality Level

form

buffered aqueous solution

composition

Protein, ≥2.5 mg/mL biuret

extent of labeling

≥3 mol biotin per mol lectin

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Lectin from Dolichos biflorus (horse gram) is a heterotetrameric glycoprotein with a molecular mass of around 113 kDa. Dolichos biflorus agglutinin (DBA) is specific for terminal α-linked N-acetyl-D-galactosamine (GalNAc). It is a dietary lectin.

Application

Lectin from Dolichos biflorus (horse gram) has been used:
  • to characterize mouse uterine natural killer (uNK) cells
  • in double staining with lectins to localize nuclear factor-κB (NF-κB) activation
  • as a species-specific gonocyte marker for the determination of donor cell colonization
  • to label undifferentiated spermatogonia/cat testicular tissue

Biochem/physiol Actions

Dolichos biflorus agglutinin (DBA) possesses binding activity for which it is used as a marker binucleate cells and can indicate the state of their differentiation. It serves as a label for primordial germ cells (PGCs), gonocytes, pre-spermatogonia and spermatogonia.
DBA has anti-A1 human blood group specificity, useful for distinguishing between A1 and A2 blood types. DBA has strong anti-Cad activity, and has an affinity for terminal N-acetyl-α-D-galactosaminyl residues. Conjugates are prepared from affinity purified lectin.

Packaging

Sold on the basis of mg active conjugate.

Physical form

contains 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spermatogonial stem cell organization in felid testis as revealed by Dolichos biflorus lectin
Escada-Rebelo S, et al.
Andrology, 4(6), 1159-1168 (2016)
Rajesh Dash et al.
JACC. Cardiovascular interventions, 11(2), 195-205 (2018-01-20)
The study investigated whether a dose response exists between myocardial salvage and the depth of therapeutic hypothermia. Cardiac protection from mild hypothermia during acute myocardial infarction (AMI) has yielded equivocal clinical trial results. Rapid, deeper hypothermia may improve myocardial salvage.
B Anne Croy et al.
Methods in molecular biology (Clifton, N.J.), 612, 465-503 (2009-12-25)
The term uterine natural killer (uNK) cell is applied in mice to an abundant but transient NK cell population that undergoes unique, terminal differentiation within embryo implantation sites during endometrial decidualization and pregnancy. In mice, decidualization is induced by attachment
Tubular NF-kB and AP-1 activation in human proteinuric renal disease
Mezzano SA, et al.
Kidney International, 60(4), 1366-1377 (2001)
Bovine sertoli cells colonize and form tubules in murine hosts following transplantation and grafting procedures
Zhang Z, et al.
Journal of Andrology, 29(4), 418-430 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service