Skip to Content
Merck
All Photos(1)

Key Documents

M-065

Supelco

(±)-MDEA solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

(±)-3,4-Methylenedioxyethylamphetamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO2
CAS Number:
Molecular Weight:
207.27
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

CC(CC1=CC=C(OCO2)C2=C1)NCC

InChI

1S/C12H17NO2/c1-3-13-9(2)6-10-4-5-11-12(7-10)15-8-14-11/h4-5,7,9,13H,3,6,8H2,1-2H3

InChI key

PVXVWWANJIWJOO-UHFFFAOYSA-N

General description

MDEA is an illicit recreational drug of the amphetamine and phenethylamine classes. This analytical reference standard is suitable to use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine drug testing. A 3,4-methylenedioxy derivative of N-ethylamphetamine, MDEA is abused for its psychedelic effects, similar to those of its structural analog MDMA (Ecstasy).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsadik T Abraham et al.
Journal of analytical toxicology, 33(8), 439-446 (2009-10-31)
3,4-Methylenedioxymethamphetamine (MDMA), or ecstasy, is excreted as unchanged drug, 3,4-methylenedioxyamphetamine (MDA), and free and glucuronidated/sulfated 4-hydroxy-3-methoxymethamphetamine (HMMA), and 4-hydroxy-3-methoxyamphetamine (HMA) metabolites. The aim of this paper is to describe the pattern and timeframe of excretion of MDMA and its metabolites
Markus R Meyer et al.
Drug metabolism and disposition: the biological fate of chemicals, 37(6), 1152-1156 (2009-03-21)
The 3,4-methylenedioxy-methamphetamine (MDMA)-related designer drug 3,4-methylenedioxyethylamphetamine (MDEA, Eve) is a chiral compound that is mainly metabolized by N-deethylation and demethylenation during phase I metabolism. The involvement of several cytochrome P450 (P450) isozymes in these metabolic steps has been demonstrated by
Rita Santos et al.
Journal of chemical information and modeling, 50(1), 146-154 (2009-11-11)
Active-site water molecules form an important component in biological systems, facilitating promiscuous binding or an increase in specificity and affinity. Taking water molecules into account in computational approaches to drug design or site-of-metabolism predictions is currently far from straightforward. In
A B Scholey et al.
Neuropsychobiology, 63(1), 15-21 (2010-10-22)
Our group has conducted several Internet investigations into the biobehavioural effects of self-reported recreational use of MDMA (3,4-methylenedioxymethamphetamine or Ecstasy) and other psychosocial drugs. Here we report a new study examining the relationship between self-reported Ecstasy use and traces of
Peter R Stout et al.
Journal of analytical toxicology, 34(8), 430-443 (2010-01-01)
On November 25, 2008, the U.S. Department of Health and Human Services posted a final notice in the Federal Register authorizing the use of liquid chromatography-tandem mass spectrometry (LC-MS-MS) and other technologies in federally regulated workplace drug testing (WPDT) programs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service