Skip to Content
Merck
All Photos(1)

Key Documents

M-015

Supelco

Methaqualone solution

1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14N2O
CAS Number:
Molecular Weight:
250.30
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

concentration

1 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

Cc1ccccc1N2C(C)=Nc3ccccc3C2=O

InChI

1S/C16H14N2O/c1-11-7-3-6-10-15(11)18-12(2)17-14-9-5-4-8-13(14)16(18)19/h3-10H,1-2H3

InChI key

JEYCTXHKTXCGPB-UHFFFAOYSA-N

General description

Methaqualone is a sedative-hypnotic and a central nervous system depressant sold under the trade name Quaalude for treatment of insomnia. Methaqualone also has a history of abuse, known by street names such as Disco biscuits, Lemmons, and 714s. This certified solution standard is applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from urine drug testing to forensic analysis and clinical toxicology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Klimes et al.
Die Pharmazie, 51(9), 651-653 (1996-09-01)
A HPLC method for quantification of kebuzone and its metabolites in whole blood was developed. The compounds and the internal standard were isolated from blood by solid-phase extraction on a C-18 cartridge. A blood sample was to be hemolyzed before
Mark A Gillman et al.
The International journal of neuroscience, 116(7), 859-869 (2006-07-25)
This article reports the first single-blind study using psychotropic analgesic nitrous oxide (PAN) for treating acute withdrawal states following the abuse of methaqualone combined and smoked with cannabis. Smoked methaqualone combined with cannabis is called "white pipe" (WP). South Africa
F Benoist et al.
European journal of biochemistry, 240(3), 713-720 (1996-09-15)
The role of microsomal triacylglycerol transfer protein (MTP) in the secretion of apolipoprotein B-100 (apoB-100) has been studied using an inhibitor of MTP: 4'-bromo-3'-methylmetaqualone. In vitro, this compound inhibits trioleoylglycerol transfer between lipid vesicles mediated by MTP with an IC50
S Herre et al.
Journal of chromatography. B, Biomedical sciences and applications, 692(1), 111-126 (1997-04-25)
The effect of the structural change in the metabolization of drugs on the HPLC retention time with an RP8 column with an acetonitrile-phosphate buffer (pH 2.3) as the mobile phase was investigated at model compound pairs of 29 functionalization reactions.
Bénédicte Manoury et al.
Nature immunology, 3(2), 169-174 (2002-01-29)
Little is known about the processing of putative human autoantigens and why tolerance is established to some T cell epitopes but not others. Here we show that a principal human HLA-DR2-restricted epitope--amino acids 85-99 of myelin basic protein, MBP(85-99)--contains a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service