Skip to Content
Merck
All Photos(2)

Key Documents

850467P

Avanti

18:0-18:1 PC

Avanti Research - A Croda Brand

Synonym(s):

1-octadecanoyl-2--(9Z-octadecenoyl)-sn-glycero-3-phosphocholine; SOPC; PC(18:0/18:1(9Z)).

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H86NO8P
CAS Number:
Molecular Weight:
788.13
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

description

1-stearoyl-2-oleoyl-sn-glycero-3-phosphocholine

Assay

>99% (TLC)

form

powder

packaging

pkg of 2 × 100 mg (850467P-200mg)
pkg of 1 × 25 mg (850467P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O)=O

InChI

1S/C44H86NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h21,23,42H,6-20,22,24-41H2,1-5H3/b23-21-/t42-/m1/s1

InChI key

ATHVAWFAEPLPPQ-VRDBWYNSSA-N

General description

18:0-18:1 PC or 1-stearoyl,2-oleoylphosphatidylcholine (SOPC) belongs to the subclass of naturally occurring phosphatidylcholines (PCs). It is the most abundant phospholipid in the animal membranes. SOPC contains one saturated sn-1 chain and one monounsaturated sn-2 chain.

Application

18:0-18:1 PC or 1-stearoyl,2-oleoylphosphatidylcholine has been used in preparation of lipid vesicles by an electro formation procedure. It is suitable to study its barotropic phase transitions by high-pressure optical method.

Biochem/physiol Actions

18:0-18:1 PC or 1-stearoyl,2-oleoylphosphatidylcholine (SOPC) facilitates the construction of various supported lipid bilayers.

Packaging

5 mL Clear Glass Sealed Ampule (850467P-200mg)
5 mL Clear Glass Sealed Ampule (850467P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tethered and polymer supported bilayer lipid membranes: Structure and function
Andersson J and Koper I
Membranes, 6(2), 30-30 (2016)
Compact and tunable size-based dielectrophoretic flow fractionation
Chuang HS, et al.
Journal of Micromechanics and Microengineering, 24(12), 125016-125016 (2014)
Effect of salicylate on the elasticity, bending stiffness, and strength of SOPC membranes
Zhou Y and Raphael RM
Biophysical Journal, 89(3), 1789-1801 (2005)
The effect of side-chain analogues of cholesterol on the thermotropic phase behavior of 1-stearoyl-2-oleoylphosphatidylcholine bilayers: a differential scanning calorimetric study
Vilcheze C, et al.
Biochimica et Biophysica Acta - Biomembranes, 1279(2), 235-242 (1996)
Barotropic phase transitions of dioleoylphosphatidylcholine and stearoyl-oleoylphosphatidylcholine bilayer membranes
Kaneshina S, et al.
Biochimica et Biophysica Acta - Biomembranes, 1374(1-2), 1-8 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service