Skip to Content
Merck
All Photos(1)

Key Documents

W390607

Sigma-Aldrich

2′-Aminoacetophenone

≥98%

Synonym(s):

2-Acetylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4COCH3
CAS Number:
Molecular Weight:
135.16
FEMA Number:
3906
Beilstein:
386122
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
11.008
NACRES:
NA.21
grade:
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Halal
Kosher

Assay

≥98%

refractive index

n20/D 1.614 (lit.)

bp

85-90 °C/0.5 mmHg (lit.)

density

1.112 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

grape

storage temp.

2-8°C

SMILES string

CC(=O)c1ccccc1N

InChI

1S/C8H9NO/c1-6(10)7-4-2-3-5-8(7)9/h2-5H,9H2,1H3

InChI key

GTDQGKWDWVUKTI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2′-Aminoacetophenone is one of the key volatile flavor components of masa corn flour products.{9] It is also reported to be responsible for the grape-like odor in culture media growing Pseudomonas aeruginosa.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of 2-aminoacetophenone production in identification of Pseudomonas aeruginosa.
Cox CD & Parker J.
Journal of Clinical Microbiology, 9(4), 479-484 (1979)
Andrew P Worth et al.
Methods in molecular biology (Clifton, N.J.), 930, 125-162 (2012-10-23)
In this chapter, a range of computational tools for applying QSAR and grouping/read-across methods are described, and their integrated use in the computational assessment of genotoxicity is illustrated through the application of selected tools to two case-study compounds-2-amino-9H-pyrido[2,3-b]indole (AαC) and
N Christoph et al.
Advances in experimental medicine and biology, 467, 659-669 (2000-03-18)
Tryptophan (TRP) and its metabolites are considered as potential precursors of 2-aminoacetophenone (AAP) in different food products causing different off-flavors. AAP is also responsible for the "untypical aging flavor (UTA)" in wine, developing a floor polish-like flavor in white wines
R E Page et al.
Experientia, 44(3), 270-271 (1988-03-15)
o-Aminoacetophenone is a pheromone produced by virgin honeybee queens and released with feces. In small social groups, the pheromone repels and is used to terminate agonistic interactions between queens and workers.
Katrin Hoenicke et al.
Journal of agricultural and food chemistry, 50(15), 4303-4309 (2002-07-11)
Kynurenine (1) and indole-3-acetic acid (2) are considered as potential precursors of 2-aminoacetophenone (3), which is regarded to be the aroma impact compound causing an "untypical aging off-flavor" (UTA) in Vitis vinifera wines. The mechanism of the formation of 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service