Skip to Content
Merck
All Photos(2)

Key Documents

A37804

Sigma-Aldrich

2′-Aminoacetophenone

98%

Synonym(s):

2-Acetylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4COCH3
CAS Number:
Molecular Weight:
135.16
Beilstein:
386122
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.614 (lit.)

bp

85-90 °C/0.5 mmHg (lit.)

density

1.112 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)c1ccccc1N

InChI

1S/C8H9NO/c1-6(10)7-4-2-3-5-8(7)9/h2-5H,9H2,1H3

InChI key

GTDQGKWDWVUKTI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katrin Hoenicke et al.
Journal of agricultural and food chemistry, 50(15), 4303-4309 (2002-07-11)
Kynurenine (1) and indole-3-acetic acid (2) are considered as potential precursors of 2-aminoacetophenone (3), which is regarded to be the aroma impact compound causing an "untypical aging off-flavor" (UTA) in Vitis vinifera wines. The mechanism of the formation of 3
W Y Li et al.
Journal of chromatography, 619(1), 148-153 (1993-09-08)
Mitomycin C (MMC) is used in the treatment of disseminated adenocarcinoma of the stomach and pancreas and is used in ophthalmology as adjunctive therapy in trabeculectomy. Since only small volumes of aqueous humor are available for analysis, a sensitive method
Amy Scott-Thomas et al.
Journal of breath research, 5(4), 046002-046002 (2011-06-28)
2-Aminoacetophenone can be detected in the breath of Pseudomonas aeruginosa colonized cystic fibrosis patients; however, low levels were also detected in a small proportion of healthy subjects. It was hypothesized that food, beverages, cosmetics or medications could be a source
Guogang Zhao et al.
Biotechnology letters, 26(16), 1255-1259 (2004-10-16)
A new isolate of Arthrobacter sulfureus , when incubated at 50 g resting cells (dry cell wt) l(-1) with 50 g glucose l(-1) and 1 g 2-aminoacetophenone l(-1) in 50 mm potassium buffer (pH 7, 4 ml) at 30 degrees
Madhushree Y Gokhale et al.
Journal of pharmaceutical sciences, 98(12), 4639-4649 (2009-06-25)
Glycosylation reaction kinetics of a series of aromatic amines (kynurenine, 2'-aminoacetophenone, daptomycin, and sulfamethoxazole) was compared to propose a unifying reaction mechanism. Kinetic studies were conducted in aqueous solutions containing glucose in the pH range 1-6.5 with 2'-aminoacetophenone and daptomycin.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service