Skip to Content
Merck
All Photos(1)

Key Documents

W362603

Sigma-Aldrich

Dihydro-β-ionone

≥90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22O
CAS Number:
Molecular Weight:
194.31
FEMA Number:
3626
EC Number:
Council of Europe no.:
11060
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.131
NACRES:
NA.21
Organoleptic:
fruity; earthy; woody; floral
grade:
Halal
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Halal

Assay

≥90%

refractive index

n20/D 1.481 (lit.)

mp

311 °C (lit.)

density

0.923 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; earthy; woody; floral

SMILES string

CC(=O)CCC1=C(C)CCCC1(C)C

InChI

1S/C13H22O/c1-10-6-5-9-13(3,4)12(10)8-7-11(2)14/h5-9H2,1-4H3

InChI key

QJJDNZGPQDGNDX-UHFFFAOYSA-N

Related Categories

General description

Dihydro-β-ionone is an aroma volatile that occurs in rose, boronia flower and juniper needles.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Essential oil and glycosidically bound volatile compounds from the needles of common juniper (Juniperus communis L.).
Mastelic J, et al.
Croatica Chemica Acta. Arhiv Za Kemiju, 73(2), 585-593 (2000)
Scent of roses?recent results.
Brunke EJ, et al.
Flavours and Fragrances, 7(4), 195-198 (1992)
Volatile constituents of Boronia megastigma flowers.
Davies NW & Menary RC.
Perfumer & Flavorist, 8, 3-8 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service