Skip to Content
Merck
All Photos(1)

Documents

93192

Sigma-Aldrich

α-Cetone

odorant used in allergy studies

Synonym(s):

3-Methyl-4-(2,6,6-trimethyl-2-cyclohexen-1-yl)-3-buten-2-one, α-Cetone, Isomethyl-α-ionone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22O
CAS Number:
Molecular Weight:
206.32
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.52

grade

for molecular biology

form

liquid

composition

iso-ɑ-methylionone, ≥85%
iso-β-methylionone, ≤10%

density

0.93 g/mL at 20 °C (lit.)

storage temp.

room temp

SMILES string

CC(=O)C(\C)=C\C1C(C)=CCCC1(C)C

InChI

1S/C14H22O/c1-10-7-6-8-14(4,5)13(10)9-11(2)12(3)15/h7,9,13H,6,8H2,1-5H3/b11-9+

InChI key

JRJBVWJSTHECJK-PKNBQFBNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methyl-4-(2,6,6-trimethyl-2-cyclohexen-1-yl)-3-buten-2-one is fragrant sensitizer added frequently to deodorants and perfumes. It is one of the allergens responsible for axillary dermatitis.

Application

Suitable for
  • contact sensitization studies to identify allergens present in cosmetic preparations
  • elucidation of molecular pathways related to allergic reactions
  • analytical standard

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

230.5 °F - closed cup

Flash Point(C)

110.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Valerie T Politano et al.
International journal of toxicology, 26(3), 271-276 (2007-06-15)
Alpha-iso-methylionone, a widely used fragrance ingredient, was evaluated for developmental toxicity in presumed pregnant Sprague-Dawley rats (25/group). Oral dosages of 0, 3, 10, or 30 mg/kg/day alpha-iso-methylionone in corn oil were administered by gavage on gestational days 7 to 17.
Susumu Ishizaki et al.
Bioscience, biotechnology, and biochemistry, 68(5), 1164-1166 (2004-06-01)
The main biodegradation product of (+/-)-alpha-isomethylionone (2) with standard activated sludge was characterized as (+/-)-1-(2,6,6-trimethyl-2-cyclohexen-1-yl)propan-2-one (1) by its analysis and synthesis. Both enantiomers (1a and 1b) of 1 were synthesized by starting from (R)- and (S)-2,4,4-trimethyl-2-cyclohexen-1-ol (3a and 3b), respectively.
A Lapczynski et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 45 Suppl 1, S280-S289 (2007-11-27)
A toxicologic and dermatologic review of alpha-iso-methylionone when used as a fragrance ingredient is presented.
Susun An et al.
Contact dermatitis, 53(6), 320-323 (2005-12-21)
The purpose of this study is to determine the frequency of responses to selected fragrances in patients with suspected fragrance allergy and to evaluate the risk factors. 9 dermatology departments of university hospitals have participated in this study for the
Valerie T Politano et al.
International journal of toxicology, 31(6), 595-601 (2013-01-04)
Alpha-iso-methylionone (AIM), a fragrance ingredient, was evaluated for systemic toxicity in rats. Male and female Sprague Dawley rats were administered 0, 5, 30, or 500 mg/kg/d AIM via gavage for 90 days. Statistically significant changes in blood chemistry parameters (reduced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service