Skip to Content
Merck
All Photos(1)

Documents

W322120

Sigma-Aldrich

Phenethyl hexanoate

natural (US), ≥97%, FG

Synonym(s):

Phenethyl caproate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CO2CH2CH2C6H5
CAS Number:
Molecular Weight:
220.31
FEMA Number:
3221
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.261
NACRES:
NA.21

biological source

botanical

Quality Level

grade

FG
Fragrance grade
Halal
Kosher
natural (US)

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥97%

refractive index

n20/D 1.486 (lit.)

density

0.971 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

honey; floral; woody; waxy; sweet

SMILES string

CCCCCC(=O)OCCc1ccccc1

InChI

1S/C14H20O2/c1-2-3-5-10-14(15)16-12-11-13-8-6-4-7-9-13/h4,6-9H,2-3,5,10-12H2,1H3

InChI key

BUYNWUMUDHPPDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phenethyl hexanoate is an ester found naturally in red wine, stinkhorn mushrooms, fermented leaves of Cassia obtusifolia, etc. It is used commercially as a flavor ingredient.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Constituents of Kawal, fermented Cassia obtusifolia leaves, a traditional food from Chad.
Mbaiguinam M, et al.
African Journal of Biotechnology, 4(10) (2005)
Vapor pressures and vaporization enthalpies of a series of esters used in flavors by correlation gas chromatography.
Kozlovskiy, M, et al.
The Journal of Chemical Thermodynamics, 86(10), 65-74 (2015)
Volatile compounds in stinkhorn (Phallus impudicus L. ex Pers.) at different stages of growth.
Pudil F, et al.
European Scientific Journal, 10(9) (2014)
Aroma recovery in wine dealcoholization by SCC distillation.
Belisario-Sanchez YY, et al.
Food and Bioprocess Technology, 5(6), 2529-2539 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service